MilliporeSigma
Derivatization of carbohydrates for GC and GC-MS analyses.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2010-12-28)
A I Ruiz-Matute, O Hernández-Hernández, S Rodríguez-Sánchez, M L Sanz, I Martínez-Castro
ABSTRACT
GC and GC-MS are excellent techniques for the analysis of carbohydrates; nevertheless the preparation of adequate derivatives is necessary. The different functional groups that can be found and the diversity of samples require specific methods. This review aims to collect the most important methodologies currently used, either published as new procedures or as new applications, for the analysis of carbohydrates. A high diversity of compounds with diverse functionalities has been selected: neutral carbohydrates (saccharides and polyalcohols), sugar acids, amino and iminosugars, polysaccharides, glycosides, glycoconjugates, anhydrosugars, difructose anhydrides and products resulting of Maillard reaction (osuloses, Amadori compounds). Chiral analysis has also been considered, describing the use of diastereomers and derivatives to be eluted on chiral stationary phases.
Product Description
Supelco
O-Ethylhydroxylamine hydrochloride, derivatization grade (GC derivatization), LiChropur
Pricing and availability is not currently available.