MilliporeSigma
  • Condensin minimizes topoisomerase II-mediated entanglements of DNA in vivo.
Condensin minimizes topoisomerase II-mediated entanglements of DNA in vivo.
The EMBO journal (2020-11-07)
Sílvia Dyson, Joana Segura, Belén Martínez-García, Antonio Valdés, Joaquim Roca
ABSTRACT
The juxtaposition of intracellular DNA segments, together with the DNA-passage activity of topoisomerase II, leads to the formation of DNA knots and interlinks, which jeopardize chromatin structure and gene expression. Recent studies in budding yeast have shown that some mechanism minimizes the knotting probability of intracellular DNA. Here, we tested whether this is achieved via the intrinsic capacity of topoisomerase II for simplifying the equilibrium topology of DNA; or whether it is mediated by SMC (structural maintenance of chromosomes) protein complexes like condensin or cohesin, whose capacity to extrude DNA loops could enforce dissolution of DNA knots by topoisomerase II. We show that the low knotting probability of DNA does not depend on the simplification capacity of topoisomerase II nor on the activities of cohesin or Smc5/6 complexes. However, inactivation of condensin increases the occurrence of DNA knots throughout the cell cycle. These results suggest an in vivo role for the DNA loop extrusion activity of condensin and may explain why condensin disruption produces a variety of alterations in interphase chromatin, in addition to persistent sister chromatid interlinks in mitotic chromatin.
Sigma-Aldrich
ICRF-193, apoptosis inducer, arabinosidase substrate
Pricing and availability is not currently available.