- Cytochrome c: functions beyond respiration.
Cytochrome c: functions beyond respiration.
Nature reviews. Molecular cell biology (2008-06-24)
Yong-Ling P Ow, Douglas R Green, Zhenyue Hao, Tak W Mak
PMID18568041
ABSTRACT
Cytochrome c is primarily known for its function in the mitochondria as a key participant in the life-supporting function of ATP synthesis. However, when a cell receives an apoptotic stimulus, cytochrome c is released into the cytosol and triggers programmed cell death through apoptosis. The release of cytochrome c and cytochrome-c-mediated apoptosis are controlled by multiple layers of regulation, the most prominent players being members of the B-cell lymphoma protein-2 (BCL2) family. As well as its role in canonical intrinsic apoptosis, cytochrome c amplifies signals that are generated by other apoptotic pathways and participates in certain non-apoptotic functions.
Sigma-Aldrich
Cytochrome c from bovine heart, ≥95% based on Mol. Wt. 12,327 basis, powder, suitable for mammalian cell culture
Pricing and availability is not currently available.