MilliporeSigma
  • A new trend in rhodamine-based chemosensors: application of spirolactam ring-opening to sensing ions.
A new trend in rhodamine-based chemosensors: application of spirolactam ring-opening to sensing ions.
Chemical Society reviews (2008-07-24)
Ha Na Kim, Min Hee Lee, Hyun Jung Kim, Jong Seung Kim, Juyoung Yoon
ABSTRACT
This tutorial review focuses on the recent development of rhodamine derivatives, in which the spirolactam (non-fluorescent) to ring-opened amide (fluorescent) process was utilized.