MilliporeSigma
  • A B-cell-homing chemokine made in lymphoid follicles activates Burkitt's lymphoma receptor-1.
A B-cell-homing chemokine made in lymphoid follicles activates Burkitt's lymphoma receptor-1.
Nature (1998-03-05)
M D Gunn, V N Ngo, K M Ansel, E H Ekland, J G Cyster, L T Williams
ABSTRACT
Secondary lymphoid organs (spleen, lymph nodes and Peyer's patches) are divided into compartments, such as B-cell zones (follicles) and T-cell zones, which provide specialized environments for specific steps of the immune response. Migration of lymphocyte subsets into these compartments is essential for normal immune function, yet the molecular cues guiding this cellular traffic are poorly defined. Chemokines constitute a family of chemotactic cytokines that have been shown to direct the migration of leukocytes during inflammation and which may be involved in the constitutive homing of lymphocytes into follicles and T-cell zones. Here we describe a novel chemokine, B-lymphocyte chemoattractant (BLC), that is strongly expressed in the follicles of Peyer's patches, the spleen and lymph nodes. BLC strongly attracts B lymphocytes while promoting migration of only small numbers of T cells and macrophages, and therefore is the first chemokine to be identified that is selective towards B cells. An orphan chemokine receptor, Burkitt's lymphoma receptor 1 (BLR-1), has been found to be required for B-cell migration into lymphoid follicles. We show that BLC stimulates calcium influx into, and chemotaxis of, cells transfected with BLR-1. Our results indicate that BLC functions as a BLR-1 ligand and may guide B lymphocytes to follicles in secondary lymphoid organs.
Sigma-Aldrich
B-Lymphocyte Chemoattractant human, ≥95% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Pricing and availability is not currently available.