MilliporeSigma
Heritable genome editing in C. elegans via a CRISPR-Cas9 system.
Nature methods (2013-07-03)
Ari E Friedland, Yonatan B Tzur, Kevin M Esvelt, Monica P Colaiácovo, George M Church, John A Calarco
ABSTRACT
We report the use of clustered, regularly interspaced, short palindromic repeats (CRISPR)-associated endonuclease Cas9 to target genomic sequences in the Caenorhabditis elegans germ line using single-guide RNAs that are expressed from a U6 small nuclear RNA promoter. Our results demonstrate that targeted, heritable genetic alterations can be achieved in C. elegans, providing a convenient and effective approach for generating loss-of-function mutants.
Sigma-Aldrich
CRISPR/Cas9 Products and Services, Design and order CRISPR gRNA, Cas9, screening libraries, controls and companion products. Formats include plant, lentivirus, IVT-RNA, plasmid, synthetic, and protein.
Pricing and availability is not currently available.