MilliporeSigma
  • The spectrum of aldolase B (ALDOB) mutations and the prevalence of hereditary fructose intolerance in Central Europe.
The spectrum of aldolase B (ALDOB) mutations and the prevalence of hereditary fructose intolerance in Central Europe.
Human mutation (2005-05-10)
René Santer, Johannes Rischewski, Michaela von Weihe, Marko Niederhaus, Sonja Schneppenheim, Kurt Baerlocher, Alfried Kohlschütter, Ania Muntau, Hans-Georg Posselt, Beat Steinmann, Reinhard Schneppenheim
ABSTRACT
We investigated the molecular basis of hereditary fructose intolerance (HFI) in 80 patients from 72 families by means of a PCR-based mutation screening strategy, consisting of heteroduplex analysis, restriction enzyme digest, DNA single strand electrophoresis, and direct sequencing. For a subset of patients mutation screening with DHPLC was established which turned out to be as fast and as sensitive as the more conventional methods. Fifteen different mutations of the aldolase B (ALDOB) gene were identified in HFI patients. As in smaller previous studies, p.A150P (65%), p.A175D (11%) and p.N335K (8%) were the most common mutated alleles, followed by c.360_363delCAAA, p.R60X, p.Y204X, and c.865delC. Eight novel mutations were identified in eight families with HFI: a small indel mutation (c.1044_1049delTTCTGGinsACACT), two small deletions (c.345_372del28; c.841_842delAC), two splice site mutations (c.113-1G>A, c.799+2T>A), one nonsense mutation (c.612T>G (p.Y204X)), and two missense mutations (c.532T>C (p.C178R), c.851T>C (p.L284P)). By mutation screening for the three most common ALDOB mutations by DHPLC in 2,000 randomly selected newborns we detected 21 heterozygotes. Based on these data and after correction for less common and private ALDOB mutations, HFI prevalence in central Europe is estimated to be 1:26,100 (95% confidence interval 1: 12,600-79,000).
Sigma-Aldrich
Aldolase from rabbit muscle, ammonium sulfate suspension, 10-20 units/mg protein
Pricing and availability is not currently available.
Sigma-Aldrich
Aldolase from rabbit muscle, lyophilized powder, ≥8.0 units/mg protein
Pricing and availability is not currently available.