MilliporeSigma
  • The microtubule depolymerizing agent naphthazarin induces both apoptosis and autophagy in A549 lung cancer cells.
The microtubule depolymerizing agent naphthazarin induces both apoptosis and autophagy in A549 lung cancer cells.
Apoptosis : an international journal on programmed cell death (2011-06-15)
Bipul R Acharya, Surela Bhattacharyya, Diptiman Choudhury, Gopal Chakrabarti
ABSTRACT
Naphthazarin (DHNQ, 5,8-dihydroxy-l,4-naphthoquinone) is a naturally available 1,4-naphthoquinone derivatives. In this study, we focused on elucidating the cytotoxic mechanism of naphthazarin in A549 non-small cell lung carcinoma cells. Naphthazarin reduced the A549 cell viability considerably with an IC(50) of 16.4 ± 1.6 μM. Naphthazarin induced cell death in a dose- and time-dependent manner by activating apoptosis and autophagy pathways. Specifically, we found naphthazarin inhibited the PI3K/Akt cell survival signalling pathway, measured by p53 and caspase-3 activation, and PARP cleavage. It also resulted in an increase in the ratio of Bax/Bcl2 protein levels, indicating activation of the mitochondrial apoptotic pathway. Similarly naphthazarin triggered LC3II expression and induced autophagic flux in A549 cells. We demonstrated further that naphthazarin is a microtubule inhibitor in cell-free system and in A549 cells. Naphthazarin treatment depolymerized interphase microtubules and disorganised spindle microtubules and the majority of cells arrested at the G(2)/M transition. Together, these data suggest that naphthazarin, a microtubule depolymerizer which activates dual cell death machineries, could be a potential novel chemotherapeutic agent.
Sigma-Aldrich
5,8-Dihydroxy-1,4-naphthoquinone, technical grade
Pricing and availability is not currently available.