MilliporeSigma
  • Increased abundance and transferability of resistance genes after field application of manure from sulfadiazine-treated pigs.
Increased abundance and transferability of resistance genes after field application of manure from sulfadiazine-treated pigs.
Applied and environmental microbiology (2013-01-15)
Sven Jechalke, Christoph Kopmann, Ingrid Rosendahl, Joost Groeneweg, Viola Weichelt, Ellen Krögerrecklenfort, Nikola Brandes, Mathias Nordwig, Guo-Chun Ding, Jan Siemens, Holger Heuer, Kornelia Smalla
ABSTRACT
Spreading manure containing antibiotics in agriculture is assumed to stimulate the dissemination of antibiotic resistance in soil bacterial populations. Plant roots influencing the soil environment and its microflora by exudation of growth substrates might considerably increase this effect. In this study, the effects of manure from pigs treated with sulfadiazine (SDZ), here called SDZ manure, on the abundance and transferability of sulfonamide resistance genes sul1 and sul2 in the rhizosphere of maize and grass were compared to the effects in bulk soil in a field experiment. In plots that repeatedly received SDZ manure, a significantly higher abundance of both sul genes was detected compared to that in plots where manure from untreated pigs was applied. Significantly lower abundances of sul genes relative to bacterial ribosomal genes were encountered in the rhizosphere than in bulk soil. However, in contrast to results for bulk soil, the sul gene abundance in the SDZ manure-treated rhizosphere constantly deviated from control treatments over a period of 6 weeks after manuring, suggesting ongoing antibiotic selection over this period. Transferability of sulfonamide resistance was analyzed by capturing resistance plasmids from soil communities into Escherichia coli. Increased rates of plasmid capture were observed in samples from SDZ manure-treated bulk soil and the rhizosphere of maize and grass. More than 97% of the captured plasmids belonged to the LowGC type (having low G+C content), giving further evidence for their important contribution to the environmental spread of antibiotic resistance. In conclusion, differences between bulk soil and rhizosphere need to be considered when assessing the risks associated with the spreading of antibiotic resistance.
Sigma-Aldrich
Sulfadiazine, 99.0-101.0%
Pricing and availability is not currently available.
Supelco
Sulfadiazine, VETRANAL®, analytical standard
Pricing and availability is not currently available.