MilliporeSigma
  • High-temperature liquid chromatography coupled on-line to a continuous-flow biochemical screening assay with electrospray ionization mass spectrometric detection.
High-temperature liquid chromatography coupled on-line to a continuous-flow biochemical screening assay with electrospray ionization mass spectrometric detection.
Analytical chemistry (2005-12-15)
Arjen R de Boer, Juan M Alcaide-Hidalgo, Johannes G Krabbe, Jeffrey Kolkman, Caspar N van Emde Boas, Wilfried M A Niessen, Henk Lingeman, Hubertus Irth
ABSTRACT
The potential of high-temperature liquid chromatography (HTLC) was investigated in an on-line combination with a screening system for bioactive compounds against the enzyme cathepsin B. Samples were separated by HTLC and subsequently analyzed by an on-line continuous-flow enzymatic assay. Detection was performed by electrospray ionization mass spectrometry, revealing both the bioactivity and the molecular mass of the bioactive compounds. Compared to conventional reversed-phase liquid chromatography, the amount of methanol necessary for separation could be decreased to only 10%, which improved the compatibility of LC with a biochemical assay. Sufficient preheating of the mobile phase prior to the separation and postcolumn cooling to prevent deactivation of the enzyme, even at column temperatures as high as 208 degrees C, was achieved as indicated by the reliable peak shapes obtained. The sensitivity was comparable with previously described systems operating at ambient temperatures as similar IC50 values were obtained. Exposing the inhibitors to high temperatures did not lead to thermal decomposition. The separation of inhibitors and the subsequent biochemical assay was performed either isothermally at various temperatures or by applying various temperature gradients as well as at various flow rates. The results obtained clearly show the compatibility of HTLC with an enzymatic screening assay.
Sigma-Aldrich
CA-074, ≥98% (HPLC)
Pricing and availability is not currently available.