MilliporeSigma
  • Recurrent eosinophilic panniculitis associated with Fasciola hepatica infection.
Recurrent eosinophilic panniculitis associated with Fasciola hepatica infection.
Journal of the American Academy of Dermatology (2000-04-18)
C Perez, R Vives, M Montes, S Ostiz
ABSTRACT
Eosinophilic panniculitis is characterized by a prominent infiltration of subcutaneous fat with eosinophils. We report a case of Fasciola hepatica infection presenting with eosinophilic panniculitis successfully treated with bithionol. To our knowledge, this is the first report of recurrent eosinophilic panniculitis associated with Fasciola hepatica infection.
Product Description
Supelco
Bithionol, VETRANAL®, analytical standard
Pricing and availability is not currently available.