MilliporeSigma
  • Toxoplasma gondii: inhibitory activity and encystation effect of securinine and pyrrolidine derivatives on Toxoplasma growth.
Toxoplasma gondii: inhibitory activity and encystation effect of securinine and pyrrolidine derivatives on Toxoplasma growth.
Experimental parasitology (2010-09-14)
Michael Holmes, Anna K Crater, Bhartesh Dhudshia, Avinash N Thadani, Sirinart Ananvoranich
ABSTRACT
Securinine, an alkaloid originally isolated from Securinega suffruticosa, exhibits a wide range of biological activities, including anti-malarial activity. Along with securinine, 10 pyrrolidine derivatives, generated via the retrosynthesis of (-)-securinine, were selected and tested for their inhibitory activity against Toxoplasma gondii growth in vitro. Anti-Toxoplasma activity correlated to hydrophobicity of the tested compounds. Three pyrrolidine derivatives along with securinine inhibit Toxoplasma proliferation at the micromolar range. These compounds act on parasite proliferation in different capacities, either by slowing the growth rate or inhibiting invasion of host cells. Securinine induces bradyzoite differentiation at comparable levels to treatment with alkali media in vitro.
Sigma-Aldrich
Securinine, ≥98% (HPLC)
Pricing and availability is not currently available.