MilliporeSigma
  • Incorporation of sulphonated cyclodextrins into polypyrrole: an approach for the electro-controlled delivering of neutral drugs.
Incorporation of sulphonated cyclodextrins into polypyrrole: an approach for the electro-controlled delivering of neutral drugs.
Biosensors & bioelectronics (1995-01-01)
G Bidan, C Lopez, F Mendes-Viegas, E Vieil, A Gadelle
ABSTRACT
The electro-controlled delivery of drugs based on the doping-dedoping mechanism of Electro-Conducting Polymers is restricted to charged substances acting as dopants. In order to overcome this limitation, this study presents an approach where the trapping/delivering is based on host-guest interaction. As an example of a neutral guest, the molecule N-methylphenothiazine (NMP) is encapsulated in the host, heptasulphonated beta-cyclodextrin (beta-CDSO3-), which is tailor-made to dope PPy. The original synthetic method for beta-CDSO3- is based on sulphonation of the periodated beta-CD in the phase transfer medium. As a consequence of their size and of their multicharged character, beta-CDSO3-s are fixed dopants. The stability of the beta-CDSO3- entrapment is checked by Optical Beam Deflection (mirage effect) measurements. The ionic movements associated with the switching of the beta-CDSO3- doped PPy (PPy+, beta-CDSO3-) film appear to be mainly due to cations with this technique. Cyclic voltammetry experiments confirm the entrapment of neutral NMP by simply dipping the PPy+, beta-CDSO3- film in a CH3CN solution containing NMP. Repeated electrochemical cycling of such a reservoir electrode indicates the progressive elimination of NMP from the (PPy+, beta-CDSO3- [NMP]) film.
Sigma-Aldrich
10-Methylphenothiazine, 98%
Pricing and availability is not currently available.