MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Examination of the cutaneous absorption of copper after the use of copper-containing ointments.
Examination of the cutaneous absorption of copper after the use of copper-containing ointments.
American journal of therapeutics (2004-11-16)
Robert W Gorter, Mario Butorac, Eloy Pulido Cobian
ABSTRACT
Because copper-containing ointments are frequently used in anthroposophical medicine, a phase I trial to investigate the cutaneous absorption of copper was conducted. Sixty-one volunteers were randomized [group A: 0.4% copper (I) oxide, 13 men and 18 women (19-55 years); group B: 20% elementary copper, 11 men and 19 women (18-70 years)]. The ointment was applied over a 4-week period followed by a 4-week wash-out phase. Serum and urine copper concentrations were measured by atomic absorption spectrometry and hair copper concentration by inductive coupled plasma mass spectrometry. For statistical analysis, the Student t test for related random samples was used; alpha = 0.05 was chosen for the standard error. In group A, an increase of copper in serum and scalp hair and a decrease in urine were found in the study period. The mean serum concentration in all premenopausal women using oral contraceptives was above normal. In group B, the serum copper concentration increased significantly; in urine, it decreased, and in scalp hair, it remained stable. A higher level of serum copper was found in female volunteers using hormonal contraception. Treatment with the 2 different ointments did not cause toxic irritations on the skin, and it can therefore be deduced that the appropriate application of ointment preparations containing copper in concentrations up to 20% do not present a toxic risk.
Sigma-Aldrich
Copper, rod, diam. 3.1 mm, 99.999%
Copper (S, P), BCR®, certified reference material, chips
Copper, IRMM®, certified reference material, 0.1 mm foil
Copper, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Copper, ACS reagent, granular, 10-40 mesh, ≥99.90%
Sigma-Aldrich
Copper, powder, <425 μm, 99.5% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, powder (spheroidal), 10-25 μm, 98%
Sigma-Aldrich
Copper, powder, 99.99% trace metals basis
Sigma-Aldrich
Copper, nanopowder, 40-60 nm particle size (SAXS), ≥99.5% trace metals basis
Sigma-Aldrich
Copper, nanopowder, 60-80 nm particle size (APS), ≥99.5% trace metals basis
Sigma-Aldrich
Copper, powder (dendritic), <45 μm, 99.7% trace metals basis
Sigma-Aldrich
Copper, shot, −3-+14 mesh, 99%
Copper (O), BCR®, certified reference material, rod
Continuous cast copper (O), BCR®, certified reference material, rod
Sigma-Aldrich
Copper, powder, <75 μm, 99%
Sigma-Aldrich
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Sigma-Aldrich
Copper, beads, 2-8 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, foil, ≥99.8% (complexometric)
Sigma-Aldrich
Copper, wire, diam. 1.0 mm, ≥99.9%
Sigma-Aldrich
Copper, turnings, purum p.a., ≥99.0%
Sigma-Aldrich
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis