MilliporeSigma
  • Wolbachia in two populations of Melittobia digitata Dahms (Hymenoptera: Eulophidae).
Wolbachia in two populations of Melittobia digitata Dahms (Hymenoptera: Eulophidae).
Neotropical entomology (2009-01-27)
Claudia S Copeland, Robert W Matthews, Jorge M González, Martin Aluja, John Sivinski
ABSTRACT
We investigated two populations of Melittobia digitata Dahms, a gregarious parasitoid (primarily upon a wide range of solitary bees, wasps, and flies), in search of Wolbachia infection. The first population, from Xalapa, Mexico, was originally collected from and reared on Mexican fruit fly pupae, Anastrepha ludens Loew (Diptera: Tephritidae); the other, from Athens, Georgia, was collected from and reared on prepupae of mud dauber wasps, Trypoxylon politum Say (Hymenoptera: Crabronidae). PCR studies of the ITS2 region corroborated that both parasitoid populations were the same species; this potentially provides a useful molecular taxonomic profile since females of Melittobia species are superficially similar. Amplification of the Wolbachia surface protein gene (wsp) confirmed the presence of this endosymbiont in both populations. Sequencing revealed that the Wolbachia harbored in both populations exhibited a wsp belonging to a unique subgroup (denoted here as Dig) within the B-supergroup of known wsp genes. This new subgroup of wsp may either belong to a different strain of Wolbachia from those previously found to infect Melittobia or may be the result of a recombination event. In either case, known hosts of Wolbachia with a wsp of this subgroup are only distantly related taxonomically. Reasons are advanced as to why Melittobia - an easily reared and managed parasitoid - holds promise as an instructive model organism of Wolbachia infection amenable to the investigation of Wolbachia strains among its diverse hosts.
Sigma-Aldrich
GenElute HP Plasmid Miniprep Kit, sufficient for 70 preparations
Pricing and availability is not currently available.
Sigma-Aldrich
GenElute HP Plasmid Miniprep Kit, sufficient for 350 preparations
Pricing and availability is not currently available.