MilliporeSigma
Upper-airway stimulation for obstructive sleep apnea.
The New England journal of medicine (2014-01-10)
Patrick J Strollo, Ryan J Soose, Joachim T Maurer, Nico de Vries, Jason Cornelius, Oleg Froymovich, Ronald D Hanson, Tapan A Padhya, David L Steward, M Boyd Gillespie, B Tucker Woodson, Paul H Van de Heyning, Mark G Goetting, Oliver M Vanderveken, Neil Feldman, Lennart Knaack, Kingman P Strohl
ABSTRACT
Obstructive sleep apnea is associated with considerable health risks. Although continuous positive airway pressure (CPAP) can mitigate these risks, effectiveness can be reduced by inadequate adherence to treatment. We evaluated the clinical safety and effectiveness of upper-airway stimulation at 12 months for the treatment of moderate-to-severe obstructive sleep apnea. Using a multicenter, prospective, single-group, cohort design, we surgically implanted an upper-airway stimulation device in patients with obstructive sleep apnea who had difficulty either accepting or adhering to CPAP therapy. The primary outcome measures were the apnea-hypopnea index (AHI; the number of apnea or hypopnea events per hour, with a score of ≥15 indicating moderate-to-severe apnea) and the oxygen desaturation index (ODI; the number of times per hour of sleep that the blood oxygen level drops by ≥4 percentage points from baseline). Secondary outcome measures were the Epworth Sleepiness Scale, the Functional Outcomes of Sleep Questionnaire (FOSQ), and the percentage of sleep time with the oxygen saturation less than 90%. Consecutive participants with a response were included in a randomized, controlled therapy-withdrawal trial. The study included 126 participants; 83% were men. The mean age was 54.5 years, and the mean body-mass index (the weight in kilograms divided by the square of the height in meters) was 28.4. The median AHI score at 12 months decreased 68%, from 29.3 events per hour to 9.0 events per hour (P<0.001); the ODI score decreased 70%, from 25.4 events per hour to 7.4 events per hour (P<0.001). Secondary outcome measures showed a reduction in the effects of sleep apnea and improved quality of life. In the randomized phase, the mean AHI score did not differ significantly from the 12-month score in the nonrandomized phase among the 23 participants in the therapy-maintenance group (8.9 and 7.2 events per hour, respectively); the AHI score was significantly higher (indicating more severe apnea) among the 23 participants in the therapy-withdrawal group (25.8 vs. 7.6 events per hour, P<0.001). The ODI results followed a similar pattern. The rate of procedure-related serious adverse events was less than 2%. In this uncontrolled cohort study, upper-airway stimulation led to significant improvements in objective and subjective measurements of the severity of obstructive sleep apnea. (Funded by Inspire Medical Systems; STAR ClinicalTrials.gov number, NCT01161420.).