- A convenient Negishi protocol for the synthesis of glycosylated oligo(ethynylene)s.
A convenient Negishi protocol for the synthesis of glycosylated oligo(ethynylene)s.
Organic letters (2008-09-25)
Tobias N Hoheisel, Holger Frauenrath
PMID18811175
ABSTRACT
A convenient and efficient sp-sp carbon heterocoupling protocol based on the Negishi reaction was developed, in which the required zinc diacetylide was generated from 1,4-bis(trimethylsilyl)butadiyne in situ and reacted with a bromoacetylene in apolar solvent mixtures. The method has been applied to the synthesis of unsymmetric glycosylated and symmetric diglycosylated oligo(ethynylene)s up to the octa(ethynylene).