MilliporeSigma
PEComa: another member of the MiT tumor family?
International journal of surgical pathology (2008-01-22)
Alberto Righi, Kypros Dimosthenous, Juan Rosai
ABSTRACT
We report 2 cases of PEComa, one occurring in the colon of an 11-year-old boy and the other in the bone (fibula) of a 92-year-old woman. Both tumors consisted of nests of large epithelioid cells surrounded by a fibrovascular stroma. The nuclei were large and vesicular, with prominent centrally located nucleoli. The cytoplasm was eosinophilic, with a fine to coarse granularity. Mitoses and individual cell necrosis were infrequent. Immunohistochemically, both tumors showed strong cytoplasmic expression of HMB-45 and intense nuclear positivity for TFE3. To our knowledge, nuclear positivity for TFE3 has been previously reported in only 5 cases of PEComa. Reactivity to this marker suggests that PEComa should be added to the growing list of human tumors of the so-called MiT family gene.
Sigma-Aldrich
Microphthalmia Transcription Factor (MiTF) (C5/D5) Mouse Monoclonal Antibody
Pricing and availability is not currently available.