MilliporeSigma
  • Epithelial membrane protein-2 is a novel therapeutic target in ovarian cancer.
Epithelial membrane protein-2 is a novel therapeutic target in ovarian cancer.
Clinical cancer research : an official journal of the American Association for Cancer Research (2010-07-31)
Maoyong Fu, Erin L Maresh, Robert A Soslow, Mohammad Alavi, Vei Mah, Qin Zhou, Alexia Iasonos, Lee Goodglick, Lynn K Gordon, Jonathan Braun, Madhuri Wadehra
ABSTRACT
The tetraspan protein epithelial membrane protein-2 (EMP2) has been shown to regulate the surface display and signaling from select integrin pairs, and it was recently identified as a prognostic biomarker in human endometrial cancer. In this study, we assessed the role of EMP2 in human ovarian cancer. We examined the expression of EMP2 within a population of women with ovarian cancer using tissue microarray assay technology. We evaluated the efficacy of EMP2-directed antibody therapy using a fully human recombinant bivalent antibody fragment (diabody) in vitro and ovarian cancer xenograft models in vivo. EMP2 was found to be highly expressed in >70% of serous and endometrioid ovarian tumors compared with nonmalignant ovarian epithelium using a human ovarian cancer tissue microarray. Using anti-EMP2 diabody, we evaluated the in vitro response of nine human ovarian cancer cell lines with detectable EMP2 expression. Treatment of human ovarian cancer cell lines with anti-EMP2 diabodies induced cell death and retarded cell growth, and these response rates correlated with cellular EMP2 expression. We next assessed the effects of anti-EMP2 diabodies in mice bearing xenografts from the ovarian endometrioid carcinoma cell line OVCAR5. Anti-EMP2 diabodies significantly suppressed tumor growth and induced cell death in OVCAR5 xenografts. These findings indicate that EMP2 is expressed in the majority of ovarian tumors and may be a feasible target in vivo.