MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Molecularly imprinted polymers on a silica surface for the adsorption of tobacco-specific nitrosamines in mainstream cigarette smoke.
Molecularly imprinted polymers on a silica surface for the adsorption of tobacco-specific nitrosamines in mainstream cigarette smoke.
Journal of separation science (2015-04-29)
Min-ting Li, Yong-yan Zhu, Li Li, Wen-na Wang, Yong-guan Yin, Quan-hong Zhu
ABSTRACT
Tobacco-specific nitrosamines are one of the most important groups of carcinogens in tobacco products. Using adsorbents as filter additives is an effective way to reduce tobacco-specific nitrosamines in cigarette smoke. Molecularly imprinted polymers (MIPs) using nicotinamide as template were grafted on the silica gel surface to obtain MIP@SiO2 and employed as filter additives to absorb tobacco-specific nitrosamines in mainstream cigarette smoke. Four milligrams of MIP@SiO2 per cigarette was added to the interface between filter and tobacco rod to prepare a binary filter system. The mainstream smoke was collected on an industry-standard Cambridge filter pad and extracted with ammonium acetate aqueous solution before analysis. Compared to the cigarette smoke of the control group, the levels of tobacco-specific nitrosamines with silica gel and with MIP@SiO2 were both reduced, and the adsorption rates of N-nitrosonornicotine, N-nitrosoanabasine, N-nitrosoanatabine, and 4-(methylnitrosamino)-1-(3-pyridine)-1-butanone with silica gel and with MIP@SiO2 were 20.76, 15.32, 18.79, and 18.01%, and 41.33, 34.04, 37.86, and 35.53%, respectively. Furthermore the content of total particle materials in cigarette smoke with silica gel was decreased evidently but showed no observable change with MIP@SiO2 . It indicated MIP@SiO2 could selectively reduce tobacco-specific nitrosamines in the mainstream cigarette smoke with no change to the cigarette flavor.
Sigma-Aldrich
3-(Trimethoxysilyl)propyl methacrylate, ≥97%
Sigma-Aldrich
3-(Trimethoxysilyl)propyl methacrylate, 98%
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Niacinamide, meets USP testing specifications
Sigma-Aldrich
Methyl acetoacetate, Arxada quality, ≥99% (GC)
Sigma-Aldrich
Methyl acetoacetate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Methacrylic acid, contains 250 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
N-[Tris(hydroxymethyl)methyl]acrylamide, contains ≤7% KCl, 93%
Sigma-Aldrich
Methacrylic acid, SAJ first grade, ≥98.0%
Sigma-Aldrich
Trimethylolpropane trimethacrylate, contains 250 ppm monomethyl ether hydroquinone as inhibitor, technical grade
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, SAJ first grade, ≥98.0%
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)