- Translating cell biology into therapeutic advances in Alzheimer's disease.
Translating cell biology into therapeutic advances in Alzheimer's disease.
Nature (1999-07-07)
D J Selkoe
PMID10392577
ABSTRACT
Studies of the molecular basis of Alzheimer's disease exemplify the increasingly blurred distinction between basic and applied biomedical research. The four genes so far implicated in familial Alzheimer's disease have each been shown to elevate brain levels of the self-aggregating amyloid-beta protein, leading gradually to profound neuronal and glial alteration, synaptic loss and dementia. Progress in understanding this cascade has helped to identify specific therapeutic targets and provides a model for elucidating other neurodegenerative disorders.
Sigma-Aldrich
Amyloid Precursor Protein β, Secreted human, recombinant, expressed in E. coli (N-terminal histidine tagged), solution
Pricing and availability is not currently available.