MilliporeSigma
Zinc is a novel intracellular second messenger.
The Journal of cell biology (2007-05-16)
Satoru Yamasaki, Kumiko Sakata-Sogawa, Aiko Hasegawa, Tomoyuki Suzuki, Koki Kabu, Emi Sato, Tomohiro Kurosaki, Susumu Yamashita, Makio Tokunaga, Keigo Nishida, Toshio Hirano
ABSTRACT
Zinc is an essential trace element required for enzymatic activity and for maintaining the conformation of many transcription factors; thus, zinc homeostasis is tightly regulated. Although zinc affects several signaling molecules and may act as a neurotransmitter, it remains unknown whether zinc acts as an intracellular second messenger capable of transducing extracellular stimuli into intracellular signaling events. In this study, we report that the cross-linking of the high affinity immunoglobin E receptor (Fcepsilon receptor I [FcepsilonRI]) induced a release of free zinc from the perinuclear area, including the endoplasmic reticulum in mast cells, a phenomenon we call the zinc wave. The zinc wave was dependent on calcium influx and mitogen-activated protein kinase/extracellular signal-regulated kinase kinase activation. The results suggest that the zinc wave is involved in intracellular signaling events, at least in part by modulating the duration and strength of FcepsilonRI-mediated signaling. Collectively, our findings indicate that zinc is a novel intracellular second messenger.
Sigma-Aldrich
Ammonium tetrathiomolybdate, 99.97% trace metals basis
Pricing and availability is not currently available.