- Synthesis of functionalized organotrifluoroborates via halomethyltrifluoroborates.
Synthesis of functionalized organotrifluoroborates via halomethyltrifluoroborates.
Organic letters (2006-05-05)
Gary A Molander, Jungyeob Ham
PMID16671774
ABSTRACT
[reaction: see text] Potassium bromo- and iodomethyltrifluoroborates have been prepared via in situ reaction of n-BuLi with dibromo- and diiodomethane, respectively, in the presence of trialkyl borates, followed by treatment with KHF(2). Moreover, a new synthetic method for the preparation of potassium organotrifluoroborates through nucleophilic substitution of the halide in these potassium halomethyltrifluoroborates is described.