MilliporeSigma
Efficient intracellular delivery of native proteins.
Cell (2015-04-25)
Diego S D'Astolfo, Romina J Pagliero, Anita Pras, Wouter R Karthaus, Hans Clevers, Vikram Prasad, Robert Jan Lebbink, Holger Rehmann, Niels Geijsen
ABSTRACT
Modulation of protein function is used to intervene in cellular processes but is often done indirectly by means of introducing DNA or mRNA encoding the effector protein. Thus far, direct intracellular delivery of proteins has remained challenging. We developed a method termed iTOP, for induced transduction by osmocytosis and propanebetaine, in which a combination of NaCl hypertonicity-induced macropinocytosis and a transduction compound (propanebetaine) induces the highly efficient transduction of proteins into a wide variety of primary cells. We demonstrate that iTOP is a useful tool in systems in which transient cell manipulation drives permanent cellular changes. As an example, we demonstrate that iTOP can mediate the delivery of recombinant Cas9 protein and short guide RNA, driving efficient gene targeting in a non-integrative manner.
Product Description
Roche
Cell Proliferation ELISA, BrdU (chemiluminescent)
Pricing and availability is not currently available.