MilliporeSigma
  • Phosphoglucose isomerase/autocrine motility factor mediates epithelial-mesenchymal transition regulated by miR-200 in breast cancer cells.
Phosphoglucose isomerase/autocrine motility factor mediates epithelial-mesenchymal transition regulated by miR-200 in breast cancer cells.
Cancer research (2011-03-11)
Aamir Ahmad, Amro Aboukameel, Dejuan Kong, Zhiwei Wang, Seema Sethi, Wei Chen, Fazlul H Sarkar, Avraham Raz
ABSTRACT
Phosphoglucose isomerase/autocrine motility factor (PGI/AMF) plays an important role in glycolysis and gluconeogenesis and is associated with invasion and metastasis of cancer cells. We have previously shown its role in the induction of epithelial-mesenchymal transition (EMT) in breast cancer cells, which led to increased aggressiveness; however, the molecular mechanism by which PGI/AMF regulates EMT is not known. Here we show, for the first time, that PGI/AMF overexpression led to an increase in the DNA-binding activity of NF-κB, which, in turn, led to increased expression of ZEB1/ZEB2. The microRNA-200s (miR-200s) miR-200a, miR-200b, and miR-200c are known to negatively regulate the expression of ZEB1/ZEB2, and we found that the expression of miR-200s was lost in PGI/AMF overexpressing MCF-10A cells and in highly invasive MDA-MB-231 cells, which was consistent with increased expression of ZEB1/ZEB2. Moreover, silencing of PGI/AMF expression in MDA-MB-231 cells led to overexpression of miR-200s, which was associated with reversal of EMT phenotype (i.e., mesenchymal-epithelial transition), and these findings were consistent with alterations in the relative expression of epithelial (E-cadherin) and mesenchymal (vimentin, ZEB1, ZEB2) markers and decreased aggressiveness as judged by clonogenic, motility, and invasion assays. Moreover, either reexpression of miR-200 or silencing of PGI/AMF suppressed pulmonary metastases of MDA-MB-231 cells in vivo, and anti-miR-200 treatment in vivo resulted in increased metastases. Collectively, these results suggest a role of miR-200s in PGI/AMF-induced EMT and thus approaches for upregulation of miR-200s could be a novel therapeutic strategy for the treatment of highly invasive breast cancer.
Product Description
Roche
Phosphoglucose Isomerase (PGI), from yeast
Pricing and availability is not currently available.