MilliporeSigma
  • Sialoside specificity of the siglec family assessed using novel multivalent probes: identification of potent inhibitors of myelin-associated glycoprotein.
Sialoside specificity of the siglec family assessed using novel multivalent probes: identification of potent inhibitors of myelin-associated glycoprotein.
The Journal of biological chemistry (2003-05-30)
Ola Blixt, Brian E Collins, Ingrid M van den Nieuwenhof, Paul R Crocker, James C Paulson
ABSTRACT
Ten of the 11 known human siglecs or their murine orthologs have been evaluated for their specificity for over 25 synthetic sialosides representing most of the major sequences terminating carbohydrate groups of glycoproteins and glycolipids. Analysis has been performed using a novel multivalent platform comprising biotinylated sialosides bound to a streptavidin-alkaline phosphatase conjugate. Each siglec was found to have a unique specificity for binding 16 different sialoside-streptavidin-alkaline phosphatase probes. The relative affinities of monovalent sialosides were assessed for each siglec in competitive inhibition studies. The quantitative data obtained allows a detailed analysis of each siglec for the relative importance of sialic acid and the penultimate oligosaccharide sequence on binding affinity and specificity. Most remarkable was the finding that myelin-associated glycoprotein (Siglec-4) binds with 500-10,000-fold higher affinity to a series of mono- and di-sialylated derivatives of the O-linked T-antigen (Galbeta(1-3)-GalNAc(alpha)OThr) as compared with alpha-methyl-NeuAc.
Sigma-Aldrich
Streptavidin−Alkaline Phosphatase from Streptomyces avidinii, lyophilized powder, ≥700 DEA units/mg protein
Pricing and availability is not currently available.