MilliporeSigma
  • Serum concentrations of carotenoids, retinol, and alpha-tocopherol in healthy persons determined by high-performance liquid chromatography.
Serum concentrations of carotenoids, retinol, and alpha-tocopherol in healthy persons determined by high-performance liquid chromatography.
Clinica chimica acta; international journal of clinical chemistry (1990-12-24)
Y Ito, J Ochiai, R Sasaki, S Suzuki, Y Kusuhara, Y Morimitsu, M Otani, K Aoki
ABSTRACT
Serum concentrations of alpha-carotene (AC), beta-carotene (BC), lycopene (LY), beta-cryptoxanthin (CR), zeaxanthin (including lutein. ZX), canthaxanthin (CX), retinol (RE), and alpha-tocopherol (TO) in healthy persons (618 males and 1,196 females) aged 7-86 years were determined by HPLC. Serum concentrations of BC among persons aged 20-49 years were higher with increasing age in females, but not in males. Serum CR concentrations decreased with age ranging from 7 to 39 years, while ZX concentrations rose in the age group of 20 to 59 years for both sexes. In contrast, serum RE concentrations and ratios of RE/BC and RE/CR, especially in males aged 20-49, were higher with age. Serum TO values in both sexes rose with age and were similar.