MilliporeSigma
  • What clinical characteristics and radiographic parameters are associated with patellofemoral instability after kinematically aligned total knee arthroplasty?
What clinical characteristics and radiographic parameters are associated with patellofemoral instability after kinematically aligned total knee arthroplasty?
International orthopaedics (2016-09-14)
Alexander J Nedopil, Stephen M Howell, Maury L Hull
ABSTRACT
Thirteen patients presented with patellofemoral instability out of 3212 knees treated with kinematically aligned total knee arthroplasty (KA TKA) during a nine year period. We determined the clinical characteristics and post-operative radiographic parameters associated with patellofemoral instability, and whether re-operation and patient reported outcome measures are different between patients with and without patellofemoral instability. Patients with patellofemoral instability were matched 1:3 to a control cohort based on date of surgery (±3 months), age (±10 years), sex, pre-operative knee deformity (varus or valgus), and implant brand. We analyzed clinical characteristics and seven post-operative radiographic parameters. Patellofemoral instability presented atraumatically (12 of 13) at 5 ± 4.7 months for a 0.4 % incidence at a mean follow-up of 43 ± 36 months. No pre-operative clinical characteristics were associated with instability. Patients with patellofemoral instability had greater flexion of the femoral component (11° versus 5°; p = 0.0012), a trend toward greater external rotation of the tibial component (2° versus 0°; p = 0.2704), more reoperations (9 versus 0; p = 0.0026) and a lower Oxford Knee Score (36 versus 42; p = 0.0045) than controls. Patellofemoral instability after kinematically aligned TKA is infrequent, presents atraumatically, and is associated with greater flexion of the femoral component than the control group. Minimizing flexion of the femoral component might reduce the risk of patellofemoral instability by promoting early engagement of the patella in the trochlear during knee flexion.
Sigma-Aldrich
Sigma-Sil-A, suitable for preparation of TMS derivatives
Pricing and availability is not currently available.