MilliporeSigma
Early interleukin-6 enhances hepatic ketogenesis in APP
Neurobiology of aging (2017-05-23)
Le Shi, Daina Zhao, Chen Hou, Yunhua Peng, Jing Liu, Shuangxi Zhang, Jiankang Liu, Jiangang Long
ABSTRACT
Alzheimer's disease (AD) is considered a multifactorial disease that affects the central nervous system and periphery. A decline in brain glucose metabolism is an early feature of AD and is accompanied by a phenotypic shift from aerobic glycolysis to ketogenesis. The liver is responsible for the generation of the ketone body. However, the mechanism that underlies hepatic ketogenesis in AD remains unclear. Here, we investigated hepatic ketogenesis during the early stage of AD pathogenesis in amyloid precursor protein (APP
Sigma-Aldrich
MISSION® esiRNA, targeting human RELA
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting human HMGCS2
Pricing and availability is not currently available.