MilliporeSigma
  • Intermittent Fasting Protects against Alzheimer's Disease Possible through Restoring Aquaporin-4 Polarity.
Intermittent Fasting Protects against Alzheimer's Disease Possible through Restoring Aquaporin-4 Polarity.
Frontiers in molecular neuroscience (2017-12-15)
Jingzhu Zhang, Zhipeng Zhan, Xinhui Li, Aiping Xing, Congmin Jiang, Yanqiu Chen, Wanying Shi, Li An
ABSTRACT
The impairment of amyloid-β (Aβ) clearance in the brain plays a causative role in Alzheimer's disease (AD). Polarity distribution of aquaporin-4 (AQP4) is important to remove Aβ from brain. AQP4 polarity can be influenced by the ratio of two AQP4 isoforms M1 and M23 (AQP4-M1/M23), however, it is unknown whether the ratio of AQP4-M1/M23 changes in AD. Histone deacetylase 3 has been reported to be significantly increased in AD brain. Moreover, evidence indicated that microRNA-130a (miR-130a) possibly mediates the regulation of histone deacetylase 3 on AQP4-M1/M23 ratio by repressing the transcriptional activity of AQP4-M1 in AD. This study aimed to investigate whether intermittent fasting (IF), increasing the level of an endogenous histone deacetylases inhibitor β-hydroxybutyrate, restores AQP4 polarity via miR-130a mediated reduction of AQP4-M1/M23 ratio in protection against AD. The results showed that IF ameliorated cognitive dysfunction, prevented brain from Aβ deposition, and restored the AQP4 polarity in a mouse model of AD (APP/PS1 double-transgenic mice). Additionally, IF down-regulated the expression of AQP4-M1 and histone deacetylase 3, reduced AQP4-M1/M23 ratio, and increased miR-130a expression in the cerebral cortex of APP/PS1 mice.
Sigma-Aldrich
MISSION® esiRNA, targeting human HDAC3
Pricing and availability is not currently available.