MilliporeSigma
  • Association of CD47 with natural killer cell-mediated cytotoxicity of head-and-neck squamous cell carcinoma lines.
Association of CD47 with natural killer cell-mediated cytotoxicity of head-and-neck squamous cell carcinoma lines.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2008-05-24)
Min Jung Kim, June-Chul Lee, Jae-Jung Lee, Soyeon Kim, Sang Goo Lee, Seok-Woo Park, Myung Whun Sung, Dae Seog Heo
ABSTRACT
Integrin-associated protein (CD47) binds specifically to the inhibitory receptor signal-regulatory protein. This study was designed to evaluate the role of CD47 in natural killer (NK) cell-mediated cytotoxicity against cancer cells. Head-and-neck squamous cell carcinoma (HNSCC) cell lines were analyzed for the expression of CD47 and susceptibility to NK cell-mediated killing. Cytolytic activity was assessed by (51)Cr-specific release assays and by measuring cytokine production. HNSCC cell lines that had high CD47 expression showed lower levels of NK cytotoxicity than those with low CD47 expression. After pre-treating cells with neutralizing major histocompatibility complex (MHC) class I or anti-CD47 antibodies, NK cell-mediated cytotoxicity against HNSCC cell lines increased. In addition, when CD47 cDNA was transfected into Caco-2 cells, NK cell-mediated cytotoxicity decreased. These findings suggest that CD47 may play an inhibitory role in NK cell-mediated cytotoxicity against cancer cells, implying a possible mechanism of immune escape in human cancer.