MilliporeSigma
LC/MS Analysis of Antidepressants From Urine Using an Ascentis® Express C18 Column
LC/MS Analysis of Antidepressants From Urine Using an Ascentis® Express C18 Column application for LC-MS
Materials
related product
Product No.
Description
Pricing
Citalopram hydrobromide solution
1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Duloxetine hydrochloride solution
1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Milnacipran hydrochloride solution
1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sertraline hydrochloride solution
1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Trazodone hydrochloride solution
1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Venlafaxine hydrochloride solution
1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Ascentis® Express C18, 2 μm UHPLC Column
2 μm particle size, L × I.D. 10 cm × 2.1 mm
CONDITIONS
column
Ascentis Express C18, 10cm x 2.1mm, 2.7 μm particles (53823-U)
column
Ascentis Express C18, 10cm x 2.1mm, 2.0 μm particles (50813-U)
mobile phase
[A] 5 mM ammonium acetate in 0.1% formic acid; [B] 0.1% formic acid in acetonitrile
gradient
10% B held  for 1 min; to 55% B in 10 min
flow rate
0.4 mL/min
pressure
8280 psi (570 bar)
column temp.
35 °C
detector
MS, ESI(+), MRM, m/z 247.138/71.958,  278.168/58.025, 372.054/148.011,   442.114/155.035,  298.014/43.96, 305.968/158.873, 325.128/108.95
injection
1 μL
Description
General description
Sample preparation: Prepare a stock solution with 100 ng/mL of compounds in synthetic urine (Cerilliant, part number S-020, lot 72112). Dilute this solution with 95:5 (5mM ammonium acetate in 0.1% formic acid in water: 0.1% formic acid in acetonitrile) to give a final concentration of 50 ng/mL. Filter through a Millex LH filter. Inject. Also prepared blank in same way.
Analysis Note
Seven modern antidepressants were analyzed from urine after a dilute and inject method using reversed-phase chromatography and mass spectrometric detection.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany