About This Item
Skip To
biological source
rabbit
Quality Level
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
human, rat
technique(s)
immunohistochemistry: suitable (paraffin)
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... GSC2(2928)
rat ... Gsc2(363831)
1 of 4
This Item | SAB4501328 | AV32208 | HPA043011 |
|---|---|---|---|
| biological source rabbit | biological source rabbit | biological source rabbit | biological source rabbit |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form affinity isolated antibody |
| technique(s) immunohistochemistry: suitable (paraffin), western blot: suitable | technique(s) ELISA: 1:20000, western blot: 1:500-1:1000 | technique(s) immunohistochemistry: suitable, western blot: suitable | technique(s) immunohistochemistry: 1:50- 1:200 |
| clone polyclonal | clone polyclonal | clone polyclonal | clone polyclonal |
| UniProt accession no. | UniProt accession no. | UniProt accession no. | UniProt accession no. |
General description
Immunogen
Application
Biochem/physiol Actions
Analysis Note
Human pituitary tissue lysate
Western Blotting Analysis: 0.25 µg/mL of this antibody detected Goosecoid2 in 10 µg of human pituitary tissue lysate.
Other Notes
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service


