Select a Size
About This Item
Skip To
form
liquid
Quality Level
packaging
pkg of 500 mL
manufacturer/tradename
Chemicon®
technique(s)
ELISA: suitable
immunohistochemistry: suitable
1 of 4
This Item | EPA100 | A4955 | P9209 |
|---|---|---|---|
| technique(s) ELISA: suitable, immunohistochemistry: suitable | technique(s) ELISA: suitable | technique(s) ELISA: suitable | technique(s) - |
| Quality Level 300 | Quality Level - | Quality Level 200 | Quality Level 200 |
| form liquid | form liquid | form liquid | form liquid |
| packaging pkg of 500 mL | packaging pkg of 500 mL | packaging - | packaging - |
| manufacturer/tradename Chemicon® | manufacturer/tradename Chemicon® | manufacturer/tradename - | manufacturer/tradename - |
General description
Application
Physical form
Preparation Note
Chemicon HRP conjugated antibodies may be diluted up to 1:16,000 in EPA II Buffer. Diluted conjugates are stable for at least 3 months when stored at 2° to 8°C. The buffer is suitable for direct use in E
Legal Information
Disclaimer
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
The Luminata ELISA HRP chemiluminescent substrates provide three major advantages: 1. High sensitivity for detecting as little as 0.1 pg of protein. 2. Broad dynamic detection range (up to 2,500-fold), enabling a wide range of quantitation. 3. Flexibility to adjust signal strength by one of two methods: a. Exposing the well to the luminometer for a longer time. b. Applying a more sensitive substrate to obtain signals in wells with otherwise undetectable signals.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service


