MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now
Reaction Clean-Up
A multi-channel pipette meticulously dispenses pink liquid into a transparent microplate against a backdrop of blurred lab equipment.
Cleanup of PCR reactions, restriction digests, sequencing reactions, and agarose and polyacrylamide gel extracts is essential for removing contaminants such as salts, enzymes, unincorporated dNTPs, primers, ethidium bromide, and other impurities that can interfere with downstream analysis. A selection of products for cleaning up DNA and RNA from enzymatic reactions or pre-purified samples is available for flexibility and convenience.
Products
DNA purification (7)
PCR (2)
activity assay (2)
ligand binding assay (2)
receptor binding assay (2)
DNA sequencing (1)
kits (8)
plates (3)
PCR reagents (1)
buffers (1)
Millipore (6)
Sigma-Aldrich (4)
Sort by Relevance
Showing 1-13 of 13
Change View
Showing 1-13 of 13
High Pure PCR Product Purification Kit
HPPCRPKRO
High Pure PCR Product Purification Kit
sufficient for 250 purifications (11732676001), sufficient for 50 purifications (11732668001), suitable for DNA purification
GenElute<SUP>™</SUP> PCR Clean-Up Kit
NA1020
GenElute PCR Clean-Up Kit
sufficient for 70 purifications
MultiScreen PCR<sub>384 </sub>Filter Plate
S384PCR
MultiScreen PCR384 Filter Plate
Purification of PCR products
GenElute<SUP>™</SUP>-E Single Spin DNA Cleanup Kit
EC600
GenElute-E Single Spin DNA Cleanup Kit
Reagents and consumables for clean-up of pre-purified DNA. 10, 50 or 250 purifications.
Montage SEQ<sub>96 </sub>Sequencing Reaction Cleanup Kit
LSKS096
Montage SEQ96 Sequencing Reaction Cleanup Kit
Purification of sequencing reactions
MultiScreen-PCR<sub>µ96</sub> Filter Plate
LSKMPCR
MultiScreen-PCRµ96 Filter Plate
Purification of PCR products
Montage Gel Extraction Kit
LSKGEL050
Montage Gel Extraction Kit
DNA Gel Extraction
MultiScreen-PCR<sub>96 </sub>Filter Plate
MSNU030
MultiScreen-PCR96 Filter Plate
Purification of PCR products
GenElute<SUP>™</SUP>-E Single Spin RNA Cleanup Kit
EC800
GenElute-E Single Spin RNA Cleanup Kit
Reagents and consumables for organic solvent clean-up from RNA. 10, 50 or 250 purifications.
GenElute<SUP>™</SUP>-E Organic Solvent DNA Cleanup Kit
EC700
GenElute-E Organic Solvent DNA Cleanup Kit
Reagents and consumables for organic solvent clean-up from DNA. 10, 50 or 250 purifications.
GenElute<SUP>™</SUP> 96 Well PCR Clean-Up Kit
PCR9604
GenElute 96 Well PCR Clean-Up Kit
4 x 96 well plate, sufficient for 4 96-well plate purifications
GenElute<SUP>™</SUP>-E RNA Gel Loading Buffer
EC222
GenElute-E RNA Gel Loading Buffer
2X Loading dye for RNA gel electrophoresis. 500µL for 100 samples.
MultiScreen SEQ<sub>384</sub> Filter Plate
S384SEQ
MultiScreen SEQ384 Filter Plate
Dye terminator removal
You have viewed 1-13 of 13 results
Page 1 of 1
Page 1 of 1
GenElute™-E Clean-up Kits
GenElute™-E DNA purification kits use a negative chromatography technique dependent on size exclusion to isolate and purify nucleic acid in a single spin.
  • Simplified workflow
  • Reduced plastic waste and no hazardous binding or wash buffers
  • Eco-friendly packaging
  • Portfolio includes both DNA and RNA cleanup kits
GenElute™ PCR Clean-Up Kits
  • The GenElute™ PCR Clean-Up Kit is designed for rapid purification of single-stranded or double-stranded PCR amplification products. This kit removes 99% of primers and other components to purify up to 100 μL or 10 μg of PCR amplified DNA in 8 minutes, with recoveries up to 95%.
  • The GenElute™ 96 Well PCR Clean-Up Kit allows for high throughput purification of PCR products by vacuum or centrifugation. This kit includes the necessary reagents for purification of highly pure PCR products. DNA recovery is 75-90% for fragments of 100 to 10,000 bp with removal of primers, primer-dimers, nucleotides, salts, and polymerase.
Multiscreen® PCR cleanup filter plates
Multiscreen® PCR filter plates offer fast, automatable solutions for high-throughput PCR purification, providing high purity and high recovery.
  • >99.5% primer removal
  • MultiScreen® PCR96 filter plates for 150–300 µL volume samples
  • MultiScreen® PCR384 filter plates for 20–100 µL sample volumes
  • MultiScreen® PCRµ96 microwell filter plate recommended for small fragments (1 – 150 bp) and smaller volumes (20 – 150 µL)
GenElute™ Gel Extraction Kit
The GenElute™ Gel Extraction Kit is designed for the rapid purification of linear and plasmid DNA fragments from standard or low-melting agarose gels. This kit can also be used to purify DNA from polyacrylamide gels.
  • Binds up to 10 μg of DNA
  • Recoveries up to 80%
  • Up to 3.5 g of agarose can be processed per column
  • Compatible with both standard and low-melting agarose in TAE or TBE buffer
Montage DNA Gel Extraction Kit
The Montage DNA Gel Extraction Kit is a fast, effective solution for fully functional DNA recovery from agarose gel slices. In one 10-minute spin, the agarose gel containing the DNA of interest is fragmented and compressed to extrude the DNA ready for sequencing or cloning.
  • Maximizes DNA recovery from agarose gel slices
  • One 10-minute spin recovers fully functional DNA
  • Purified DNA is ready for cloning or sequencing
SigmaSpin® Sequencing Reaction Cleanup Columns and Plates
  • SigmaSpin® Post-Reaction Clean-Up columns are ideal for lower throughput applications and provide a fast, simple, and highly efficient method for removing unincorporated dyes, excess salts, and other interfering reaction components.
  • SigmaSpin™ 96-Well Post-Reaction Clean-Up plates provide a fast, simple, and highly efficient method for removing unincorporated dyes, excess salts, and other interfering reaction components.
Montage and Multiscreen® Sequencing Reaction Cleanup Plates
Montage and MultiScreen® Sequencing Reaction Cleanup products incorporate a patented size-exclusion membrane to yield highly purified sequencing reaction products. Available in 96- and 384-well formats, the plates follow a vacuum-driven protocol, and are automation compatible.
  • 10-minute vacuum-based protocol
  • Compatible with a variety of templates
  • Optimized for use with BigDye™ chemistries