MilliporeSigma
mAbs Downstream
Products for Development and Manufacturing
Higher cell densities and antibody titers from increasingly advanced upstream processes result in complex downstream purification challenges. The focus of downstream bioprocessing is efficient recovery and purification while controlling bioburden and assuring viral safety to provide drug safety for patients. The downstream workflow, which includes clarification, chromatography, concentration, viral clearance, and sterile filtration steps, is essential for ensuring monoclonal antibody (mAb) product quality, yield, safety, and sterility. 
Prefiltration & Bioburden Control
Particulate and bioburden contaminants can cause premature fouling of aseptic filters and interfere with downstream purification. Prefilters remove particulate impurities such as colloids, aggregated and non-product proteins, lipids, and cellular debris. They also provide a cost-effective option for improving process efficiency by extending the capacity of costly sterilizing filters and reducing the impact of particulate contaminants in chromatographic separation.
Bioburden reduction filters remove particulate impurities and different types of bioburden. Downstream operations operate as “low bioburden” or “bioburden-controlled,” where bioburden is present but controlled and routinely monitored. Bioburden control strategies include assessing bioburden, mitigating the risk of occurrence, and monitoring to assure process control.
Our integrated offering of prefilters and bioburden reduction filters includes:
  • Millipore® Filters  
  • Millipore® Integrity Testers
  • Millipore® Equipment and Systems (housings)
  • Millipore® Sterile Sampling
  • Millipore® Services (single-use and filter validation, equipment installation, system qualification, system training, system services)
  • Millipore® Automation and Analytics Software
Related Materials
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
Related Categories
Four containers are depicted in the image related to buffers and pH adjusters. The two smaller containers on the left have a red lid and white body. The two larger containers on the right are blue with red-rimmed lids.
Buffers & pH Adjusters
Our broad portfolio of buffer materials manufactured under appropriate controls is tailored to your needs.
Learn More
An industrial clarification filter system featuring multiple cylindrical units mounted on a metal frame, complete with gauges and valves, designed for large-scale filtration processes.
Clarification & Harvest
Clarification steps prepare your bioreactor harvest material for downstream purification by reducing the levels of impurities and particulates.
Learn More
Chromatograph system for bioprocessing, featuring various tubes, dials, and components, predominantly silver with blue accents and a logo featuring the letter ‘M’ on the side panel.
Chromatography
Downstream purification in bioprocessing includes one or more chromatographic separation steps to purify your molecule and remove impurities.
Learn More
A female scientist wearing protective goggles holds up a sterile drug product.
Sterile Filtration and Bioburden Control
Our broad portfolio of membrane filters offers options for efficient processing of every pharma and biopharma stream.
Learn More
Dropper transferring liquid in monoclonal antibody process
Viral Clearance
Our viral clearance portfolio includes the virus filtration solutions, chemicals, holders and systems.
Learn More
Mobius® FlexReady Solution for TFF
Ultrafiltration & Diafiltration
Ultrafiltration & diafiltration increase capacity, concentration, and recovery in bioprocessing.
Learn More
Related Resources
Explore Our Solutions