Skip to Content
MilliporeSigma

12307C

Fetal Bovine Serum

suitable for cell culture, non-USA origin, USDA approved

Synonym(s):

FBS, FCS, sera, serum

Sign In to View Organizational & Contract Pricing.

Select a Size

Pricing and availability is not currently available. Contact Technical Service

About This Item

UNSPSC Code:
12352207
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

bovine fetus

sterility

sterile-filtered

quality

USDA approved

origin

non-USA origin

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Analysis Note

  • Endotoxin and hemoglobin tested
  • Tested for the presence of bacteria, virus, and mycoplasma
  • Triple filtered with 0.1 micron membrane under aseptic conditions

Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

SAFC Biosciences has established process parameters and controls for maximum inactivation of microbial contaminants while maintaining product integrity through its validation of the gamma irradiation process for serum products. The validation study evaluated package configurations, process temperatures, radiation dose response and radiation distribution within the packages. The study was conducted at both pilot scale (100 mL samples) and production scale (500 mL, 1000 mL, 3 L and 4 L samples).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service