Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
96 wells | Usually Ships in 4 Weeks. (Orders outside of US, please allow an additional 1-2 weeks for delivery) | $979.00 |
About This Item
sample type cell culture supernatant(s)
sample type serum
$979.00
species reactivity
mouse
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
input
sample type plasma
sample type cell culture supernatant(s)
sample type serum
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 6.2 pg/mL
standard curve range: 6.14-1500 pg/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
mouse ... Ifnz(319146)
General description
Application
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
1 of 1
This Item | |||
|---|---|---|---|
| description - | description - | description - | description - |
| Gene Information mouse ... Ifnz(319146) | Gene Information mouse ... Eng(13805) | Gene Information mouse ... Mme(17380) | Gene Information mouse ... Fas(14102) |
| species reactivity mouse | species reactivity mouse | species reactivity mouse | species reactivity mouse |
| assay range inter-assay cv: <12% | assay range inter-assay cv: <12% | assay range inter-assay cv: <12% | assay range inter-assay cv: <12% |
| input sample type plasma | input sample type serum | input sample type serum | input sample type serum |
| technique(s) ELISA: suitable | technique(s) ELISA: suitable | technique(s) ELISA: suitable | technique(s) ELISA: suitable |
| detection method colorimetric | detection method colorimetric | detection method colorimetric | detection method colorimetric |
signalword
Warning
hcodes
pcodes
Hazard Classifications
Met. Corr. 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Storage Class
10 - Combustible liquids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




