MilliporeSigma
HomeGel ElectrophoresisTAE and TBE Running Buffers Recipe & Video
TAE and TBE Running Buffers Recipe & Video
What are Tris Acetate EDTA and Tris Borate EDTA?
Tris acetate EDTA (TAE) and tris borate EDTA (TBE) are the two most common running buffers used in nucleic acid electrophoresis. As buffers, they have a fairly constant pH and are able to conduct electricity because of their concentration of hydrogen ions. These properties are necessary for gel electrophoresis during which proteins are separated by electric charge.
Preparing Running Buffers
TBE and TAE are most often mixed from their constituent parts into laboratory stock solutions. Both buffers can be purchased at the working concentration or in a powdered or concentrated format that is simply prepared via dilution.
Refer to the recipes below to prepare TAE and TBE in common stock solution concentrations. Our buffer calculator can also help you find the correct dilution (or conversion) for various buffers from stock solution to a desired molarity and volume.
Video: TAE and TBE Buffers for Gel Electrophoresis
Review common TAE and TBE buffer solution recipes and learn which running buffer to choose for your nucleic acid gel electrophoresis application. Research technology specialist Chris Lemke mixes up stock solutions and provides helpful buffer selection tips.
This video was built in collaboration with Seeding Labs, a nonprofit that connects universities and research institutes in developing countries with high-quality surplus lab equipment, training, and professional exchanges.
TAE Buffer 50x Stock Recipe
Adjust volume to 1 L.
10x TAE Recipe
For 1L of 10x solution,
1x TAE Recipe
Dilute 1:10
using ultrapure water.
TBE Buffer 10x Stock Recipe
Adjust volume to 1 L.
1x TBE Preparation
Dilute 10x concentrated TBE buffer 10-fold with ultrapure water.
The final solution should contain:
Buffer Prep Tips
  • If precipitation is present, warm to 37 °C and mix until completely dissolved prior to dilution.
  • It is recommended 1x working solutions be filtered through a 0.2 mm filter before use.
  • 1x working solutions can be used until the expiration date on packaging with storage at room temperature. Discard if buffer becomes cloudy or discolored.
TAE vs. TBE Comparison Chart
 TBE Buffer
TAE Buffer
Notes
Buffering CapacityHighLowTAE becomes exhausted during extended or repeated electrophoresis.
Migration of DNASlowFastTAE has better conductivity.
ResolutionHigh resolution of <2KB DNA fragments High resolution of >2KB DNA fragmentsTBE supports agarose cross-linkage.
Enzyme InhibitorYesNoBorate from TBE inhibits many enzymes.
CostHigherLower 
Applications
Tris-acetate-EDTA (TAE) running buffer and tris-borate-EDTA (TBE) are commonly used buffers for DNA agarose gel electrophoresis that are especially useful in preparative work.1
Compared to tris-borate-EDTA (TBE) and tris-phosphate-EDTA (TPE) buffers, double-stranded DNA tends to run faster in TAE. However, because TAE has the lowest buffering capacity of the three buffers, the buffering capacity can become exhausted during extended electrophoresis. Buffer circulation or replacement can remedy this situation.
TAE
The 1x TAE buffer is used both in the agarose gel and as a running buffer. Applied voltages of < 5 V/cm (the distance between the electrodes of the unit) are recommended for maximum resolution.2
TAE buffer has been utilized in agarose gel electrophoresis of RNA.3,4
A study of free DNA solution mobility in TAE at various buffer concentrations, in the presence and absence of added NaCl, has been reported.5
The use of TAE buffer in a denaturing gradient gel electrophoresis method for broad-range mutation analysis has been described.
TBE
TBE buffer is recommended for resolution of RNA and DNA fragments smaller than 1500 bp.
TBE is used with both non-denaturing or denaturing (7 M urea) gels.
It is also routinely used for DNA automated sequencing gel.
Tris-borate-EDTA buffer has been used for pulsed-field gel electrophoresis (PFGE). Applied voltages of less than 5 V/cm are recommended for maximum resolution.
Bionic Buffer is a unique alternative to traditional TBE (tris-borate-EDTA) and TAE (tris-acetate-EDTA) electrophoresis buffers. Bionic Buffer allows for:
  • Band resolution in minutes
  • Running gels 2-3x faster than in TBE/TAE
  • Sharper bands in less time
  • Use with pre-cast gels
Materials
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
References
1.
Ogden RC, Adams DA. 1987. [8] Electrophoresis in agarose and acrylamide gels.61-87. https://doi.org/10.1016/0076-6879(87)52011-0
2.
Sambrook, J, Russell, DW. 2001. Molecular Cloning: A Laboratory Manual, . 3rd ed., pp. 5.8, 5.76, A1.16.. CSHL Press (Cold Spring Harbor, NY), .
3.
Loening U. 1967. The fractionation of high-molecular-weight ribonucleic acid by polyacrylamide-gel electrophoresis. 102(1):251-257. https://doi.org/10.1042/bj1020251
4.
Masters DB. 1992. High sensitivity quantification of RNA from gels and autoradiograms with affordable optical scanning. Biotechniques. 12(6):902-906, 908-911.
5.
Stellwagen E, Stellwagen N. 2002. The free solution mobility of DNA in Tris-acetate-EDTA buffers of different concentrations, with and without added NaCl. Electrophoresis. 23 (12):1935-1941.
6.
Hayes V. 1999. Improvements in gel composition and electrophoretic conditions for broad-range mutation analysis by denaturing gradient gel electrophoresis. 27(20):29e-29. https://doi.org/10.1093/nar/27.20.e29