Repeating Unit |
Tg (°C) |
Tm (°C) |
Repeating Unit |
Tg (°C) |
Tm (°C) |
Acenaphthylene |
214 |
|
N,N-Dimethylacrylamide |
89 |
|
Acetaldehyde |
-32 |
165 |
Dimethylaminoethyl methacrylate |
19 |
|
4-Acetoxystyrene |
116 |
|
2,6-Dimethyl-1,4-phenylene oxide |
167 |
|
Acrylamide |
165 |
|
Dimethylsiloxane |
-127 |
-40 |
Acrylic acid |
105 |
|
2,4-Dimethylstyrene |
112 |
|
Acrylonitrile, syndiotactic |
125 |
319 |
2,5-Dimethylstyrene |
143 |
|
Allyl glycidyl ether |
-78 |
|
3,5-Dimethylstyrene |
104 |
|
Benzyl acrylate |
6 |
|
Dodecyl acrylate |
-3 |
|
Benzyl methacrylate |
54 |
|
Dodecyl methacrylate |
-65 |
|
Bisphenol A-alt-epichlorohydrin |
100 |
|
Dodecyl vinyl ether |
-62 |
|
Bisphenol A terephthalate |
205 |
|
Epibromohydrin |
-14 |
|
Bisphenol carbonate |
174 |
|
Epichlorohydrin |
-22 |
|
Bisphenol F carbonate |
147 |
|
1,2-Epoxybutane |
-70 |
|
Bisphenol Z carbonate |
175 |
|
1,2-Epoxydecane |
-70 |
|
4-Bromostyrene |
118 |
|
1,2-Epoxyoctane |
-67 |
|
cis-Butadiene |
102 |
1 |
2-Ethoxyethyl acrylate |
-50 |
|
trans-Butadiene |
-58 |
148 |
4-Ethoxystyrene |
86 |
|
1-Butene |
-24 |
171 |
Ethyl acrylate |
-24 |
|
N-tert-Butylacrylamide |
128 |
|
Ethyl cellulose |
43 |
|
Butyl acrylate |
-54 |
|
Ethylene, HDPE |
-125 |
130 |
sec-Butyl acrylate |
-26 |
|
Ethylene adipate |
-45 |
54 |
tert-Butyl acrylate |
43-107 |
193 |
Ethylene-trans-1,4-cyclohexyldicarboxylate |
18 |
– |
2-tert-Butylaminoethyl methacrylate |
33 |
|
Ethylene isophthatate |
51 |
|
Butyl glydicyl ether |
-79 |
|
Ethylene malonate |
-29 |
|
Butyl methacrylate |
20 |
|
Ethylene 2,6-napthalenedicarboxylate |
113 |
|
tert-Butyl methacrylate |
118 |
|
Ethylene oxide |
-66 |
66 |
4-tert-Butylstyrene |
127 |
|
Ethylene terephthalate |
72 |
265 |
tert-Butyl vinyl ether |
88 |
250 |
2-Ethylhexyl acrylate |
-50 |
|
Butyl vinyl ether |
-55 |
64 |
2-Ethylhexyl methacrylate |
-10 |
|
ε-Caprolactone |
-60 |
|
2-Ethylhexyl vinyl ether |
-66 |
|
Cellulose nitrate |
53 |
|
Ethyl methacrylate |
65 |
|
Cellulose tripropionate |
|
|
Ethyl vinyl ether |
-43 |
86 |
cis-Chlorobutadiene |
-20 |
80 |
4-Fluorostyrene |
95
|
|
trans-Chlorobutadiene |
-40 |
101 |
Formaldehyde |
-82 |
181 |
2-Chlorostyrene |
119 |
|
Hexadecyl acrylate |
35 |
|
3-Chlorostyrene |
90 |
|
Hexadecyl methacrylate |
15 |
|
4-Chlorostyrene |
110 |
|
Hexyl acrylate |
-57 |
|
Chlorotrifluoroethylene |
52 |
214 |
Hexyl methacrylate |
-5 |
|
2-Cyanoethyl acrylate |
4 |
|
2-Hydropropyl methacrylate |
76 |
|
Cyclohexyl acrylate |
19 |
|
Hydroquinone-alt-epichlorohydrin |
60 |
|
Cyclohexyl methacrylate |
92 |
|
2-Hydroxyethyl methacrylate |
57 |
|
Cyclohexyl vinyl ether |
81 |
|
Indene |
85 |
|
2,6-Dichlorostyrene |
167 |
|
Isobornyl acrylate |
94 |
|
Diethylaminoethyl methacrylate |
20 |
|
Isobornyl methacrylate |
110 |
|
Isobutyl acrylate |
-24 |
|
p-Phenylene terephthalamide |
345 |
|
Isobutylene |
-73 |
|
Phenylene vinylene |
80 |
380 |
Isobutyl methacrylate |
53 |
|
Phenyl methacrylate |
110 |
|
Isobutyl vinyl ether |
-19 |
165 |
Phenyl vinyl ketone |
74 |
|
cis-Isoprene |
-63 |
28 |
Potassium acrylate |
194 |
|
trans-Isoprene |
-66 |
65 |
Propylene, atactic |
-13 |
|
N-Isopropylacrylamide |
85-130 |
|
Propylene, isotactic |
-8 |
176 |
Isopropyl acrylate, isotactic |
-11 |
162 |
Propylene, syndiotactic |
-8 |
|
Isopropyl methacrylate |
81 |
|
Propylene oxide |
-75 |
66 |
Methacrylic acid |
228 |
|
Propyl vinyl ether |
-49 |
76 |
Methacrylic anhydride |
159 |
|
Sodium acrylate |
230 |
|
Methacrylonitrile |
120 |
|
Sodium methacrylate |
310 |
|
2-Methoxyethyl acrylate |
-50 |
|
Styrene, atactic |
100 |
|
4-Methoxystyrene |
113 |
|
Styrene, isotactic |
100 |
240 |
Methyl acrylate |
10 |
|
Tetrabromobisphenol A carbonate |
157 |
|
Methyl cellulose |
|
|
Tetrafluoroethylene |
117 |
327 |
Methyl glycidyl ether |
-62 |
|
Tetrahydrofuran |
-84 |
|
Methyl methacrylate, atactic |
105,120 |
|
Tetramethylene adipate |
-118 |
|
Methyl methacrylate, syndiotactic |
115 |
200 |
Tetramethylene terephthalate |
17 |
232 |
4-Methylpentene |
29 |
250 |
Thio-1,4-phenylene |
97 |
285 |
Methylphenylsiloxane |
-86 |
|
2,2,2-Trifluoroethyl acrylate |
-10 |
|
Methylstyrene |
20 |
|
Trimethylene oxide |
-78 |
|
3-Methylstyrene |
97 |
|
Trimethylsilyl methacrylate |
68 |
|
4-Methylstyrene |
97 |
|
2,4,6-Trimethylstyrene |
162 |
|
Methyl vinyl ether |
-31 |
144 |
Vinyl acetal |
355 |
82 |
Nylon 4,6 (tetramethylene adipamide) |
43 |
|
Vinyl acetate |
30 |
|
Nylon 6 (-caprolactam) |
52 |
225 |
Vinyl alcohol |
85 |
220 |
Nylon 6,6 (hexamethylene adipamide) |
50 |
265 |
Vinyl benzoate |
71 |
|
Nylon 6,9 (hexamethylene azelamide) |
58 |
|
Vinyl 4-tert-butylbenzoate |
101 |
– |
Nylon 6,10 (hexamethylene sebacamide) |
50 |
227 |
Vinyl butyral |
322 |
49 |
Nylon 6,12 (hexamethylene dodecane-diamide) |
46 |
|
Vinyl carbazole |
227 |
320 |
Nylon 11 (ω−undecanamide) |
42 |
189 |
Vinyl chloride |
81 |
227 |
Nylon 12 (ω-dodecanamide) |
41 |
179 |
Vinyl cyclohexanoate |
76 |
|
1-Octadecene |
55 |
|
Vinylferrocene |
189 |
|
Octadecyl methacrylate |
-100 |
|
Vinyl fluoride |
41
|
200 |
1-Octene |
-63 |
|
Vinyl formal |
105 |
|
Octyl methacrylate |
-20 |
|
Vinylidene chloride |
-18 |
200 |
Oxy-4,4'-biphenyleneoxy-1,4-phenylenesulfonyl-1,4-phenylene |
230 |
290 |
Vinylidene fluoride |
-40 |
171 |
Oxy-1,4-phenylenesulfonyl-1,4-phenyleneoxy-1,4-phenyleneisopropylidene-1,4-phenylene |
165 |
190 |
2-Vinyl naphthalene |
151 |
|
Oxy-1,4-phenylenesulfonyl-1,4-phenylene ether |
214 |
230 |
Vinyl pivalate |
86 |
|
p-Phenylene isophthalamide |
225 |
380 |
Vinyl propionate |
10 |
|
2-Vinylpyridine |
104 |
|
4-Vinylpyridine |
142 |
|
1-Vinyl-2-pyrrolidone |
54 |
|
Vinyl trifluoroacetate |
46 |
|