[Skip to Content](https://www.sigmaaldrich.com#main-content)
[](https://www.sigmaaldrich.com/US/en)
Products
Cart0
USEN
Products
ProductsApplicationsServicesResourcesSupport
[Login / Register](https://www.sigmaaldrich.com/oidc-sign-in)
[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)
[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)
Cart0
[Home](https://www.sigmaaldrich.com/US/en)Chiral Disulfonimides for Asymmetric Counteranion-directed Catalysis (ACDC)
# Chiral Disulfonimides for Asymmetric Counteranion-directed Catalysis (ACDC)
## Introduction
Asymmetric counteranion-directed catalysis (ACDC) has a number of valuable applications in enantioselective synthesis.1 Beginning in 2006, Benjamin List2 and others have expanded this field to encompass a broad range of useful transformations.
## Representative Applications
In 2009, List and coworkers reported that disulfonimide catalyst [790753](https://www.sigmaaldrich.com/US/en/product/aldrich/790753) can be used to catalyze asymmetric Mukaiyama aldol reactions in up to 94% ee.3 Two years later, this reaction was extended to vinylogous and bisvinylogous reactions as well.4
More recently, 790753 was employed as a Brønsted acid catalyst in the asymmetric Friedel-Crafts alkylation of imines with indoles.5
Finally, disulfonamide catalyst [790842](https://www.sigmaaldrich.com/US/en/product/aldrich/790842) was successfully employed in the three-component synthesis of homoallylic amines in up to 97% ee.6
To learn more about other products in our catalog that have been established in collaboration Prof. Dr. List, please visit his [Professor Product Page](https://www.sigmaaldrich.com/US/en/collections/professor-product-portal/list).
## Materials
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
### References
1\.
Mahlau M, List B. 2013. Asymmetric Counteranion-Directed Catalysis: Concept, Definition, and Applications. Angew. Chem. Int. Ed.. 52(2):518-533. [https://doi.org/10.1002/anie.201205343](https://doi.org/10.1002/anie.201205343)
2\.
Mayer S, List B. 2006. Asymmetric Counteranion-Directed Catalysis. Angew. Chem. Int. Ed.. 45(25):4193-4195. [https://doi.org/10.1002/anie.200600512](https://doi.org/10.1002/anie.200600512)
3\.
García-García P, Lay F, García-García P, Rabalakos C, List B. 2009. A Powerful Chiral Counteranion Motif for Asymmetric Catalysis. Angew. Chem. Int. Ed.. 48(24):4363-4366. [https://doi.org/10.1002/anie.200901768](https://doi.org/10.1002/anie.200901768)
4\.
Ratjen L, García-García P, Lay F, Beck ME, List B. 2011. Disulfonimide-Catalyzed Asymmetric Vinylogous and Bisvinylogous Mukaiyama Aldol Reactions. Angew. Chem. Int. Ed.. 50(3):754-758. [https://doi.org/10.1002/anie.201005954](https://doi.org/10.1002/anie.201005954)
5\.
Chen L, He H, Chan W, Lee AWM. 2011. Chiral Sulfonimide as a Brønsted Acid Organocatalyst for Asymmetric Friedel?Crafts Alkylation of Indoles with Imines. J. Org. Chem.. 76(17):7141-7147. [https://doi.org/10.1021/jo2011335](https://doi.org/10.1021/jo2011335)
6\.
Gandhi S, List B. 2013. Catalytic Asymmetric Three-Component Synthesis of Homoallylic Amines. Angew. Chem. Int. Ed.. 52(9):2573-2576. [https://doi.org/10.1002/anie.201209776](https://doi.org/10.1002/anie.201209776)
__Related Articles__
- [Jørgensen’s Organocatalysts](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/ch-functionalization/jorgensen-organocatalysts)
- [MCAT- 53™ Catalyst for Ruthenium Formation](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/ch-functionalization/mcat-53-green-ruthenium-catalyst)
- [Role of High Purity Metal Salts in Chemical Synthesis](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/ch-functionalization/role-of-high-purity-metal-salts-in-chemical-synthesis)
- [Reductive amination with 2-picoline-borane complex](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/cross-coupling/2-picoline-borane)
- [2nd Generation Buchwald Precatalysts](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/cross-coupling/2nd-generation-buchwald-precatalysts)
- [MeOBIPHEP](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/cross-coupling/MeOBIPHEP)
- [An Introduction to A-Phos, its Applications and Advantages](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/cross-coupling/a-phos)
- [Trialkylphosphine for MBH Reactions](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/cross-coupling/air-stable-trialkylphosphine)
- [View More](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search)
__Related Product Categories__
- [Chiral Catalysts & Ligands](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/catalysts/chiral-catalysts-and-ligands)
Top
__Sign In To Continue__
Sign In__Don't Have An Account?__Register
Support[Customer Support](https://www.sigmaaldrich.com/US/en/support/customer-support)[Contact Us](https://www.sigmaaldrich.com/US/en/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Safety Data Sheets (SDS)](https://www.sigmaaldrich.com/US/en/documents-search?tab=sds)[Certificates (COA/COO)](https://www.sigmaaldrich.com/US/en/documents-search?tab=coa)[Quality & Regulatory](https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management)[Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps)[Webinars](https://www.sigmaaldrich.com/US/en/collections/webinars)
Orders[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)[Custom Products](https://www.sigmaaldrich.com/US/en/services/custom-products)[eCommerce Solutions](https://www.sigmaaldrich.com/US/en/life-science/ecommerce/ecommerce-solutions)
Company[About Us](https://www.sigmaaldrich.com/US/en/life-science/about-us)[Responsibility](https://www.sigmaaldrich.com/US/en/life-science/ssbi)[Events](https://www.sigmaaldrich.com/US/en/collections/events)[Press Releases](https://www.sigmaaldrich.com/US/en/collections/press)[Programs](https://www.sigmaaldrich.com/US/en/life-science/partnership-programs)[Careers](https://careers.merckgroup.com/global/en)[Offices](https://www.sigmaaldrich.com/US/en/collections/offices)
Social Media
[](https://www.linkedin.com/company/merck-life-science)
[](https://x.com/Merck_lifesci)
[](https://www.facebook.com/Merck.lifescience)
[](https://www.instagram.com/mercklifescience/)
[](https://www.youtube.com/@MerckLifeScience/featured)
MilliporeSigma
__Research. Development. Production.__
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
[](https://www.sigmaaldrich.com/US/en)
[Sigma-Aldrich® Solutions](https://www.sigmaaldrich.com/US/en/life-science/sigma-aldrich)[BioReliance® Solutions](https://www.sigmaaldrich.com/US/en/life-science/bioreliance)[Millipore® Solutions](https://www.sigmaaldrich.com/US/en/life-science/millipore)[SAFC® Solutions](https://www.sigmaaldrich.com/US/en/life-science/safc)[Milli-Q® Solutions](https://www.sigmaaldrich.com/US/en/life-science/milliq)[Supelco® Solutions](https://www.sigmaaldrich.com/US/en/life-science/supelco)
© 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies.
Reproduction of any materials from the site is strictly forbidden without permission.
[Site Use Terms](https://www.sigmaaldrich.com/US/en/life-science/legal/site-use-terms)|[Privacy Policy](https://www.sigmaaldrich.com/US/en/life-science/legal/privacy-statement)|[General Terms and Conditions of Sale](https://www.sigmaaldrich.com/US/en/life-science/legal/terms-and-conditions)|[Copyright Consent](https://www.sigmaaldrich.com/US/en/life-science/legal/copyright-consent) |[Site Map](https://www.sigmaaldrich.com/US/en/site-map)
An unknown error has occured.