[Skip to Content](https://www.sigmaaldrich.com#main-content)
[](https://www.sigmaaldrich.com/US/en)
Products
Cart0
USEN
Products
ProductsApplicationsServicesResourcesSupport
[Login / Register](https://www.sigmaaldrich.com/oidc-sign-in)
[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)
[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)
Cart0
[Home](https://www.sigmaaldrich.com/US/en)[Fluorination](https://www.sigmaaldrich.com/US/en/applications/chemistry-and-synthesis/synthetic-methods/fluorination)AISF: A Solid Alternative to Sulfuryl Fluoride Gas
# AISF: A Solid Alternative to Sulfuryl Fluoride Gas
Fluorosulfates and sulfamoyl fluorides have applications ranging from chemical biology to polymer chemistry in addition to being valuable synthetic precursors to diaryl sulfates, sulfamides and sulfamates. Synthetic methods to construct fluorosulfates and sulfamoyl fluorides have typically relied on the use of sulfuryl fluoride gas. However, the specialized equipment and additional safety precautions required for working with a toxic gas, in addition to challenges with procurement, have precluded the broad usage of sulfuryl fluoride gas.
A solid and bench-stable alternative to sulfuryl fluoride gas has been developed by Pfizer and BioDuro chemists, 4-(acetylamino)phenyl]imidodisulfuryl difluoride (AISF, [901243](https://www.sigmaaldrich.com/US/en/product/aldrich/901243)). AISF is a shelf-stable, crystalline reagent for the installation of the valuable SO2F functional group.
![AISF - \[4-(Acetylamino)phenyl\]ImidodiSulfuryl](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/technical-documents/articles/chemistry-and-synthesis/fluorination/4-acetylamino-phenyl-Imidodi-sulfuryl.jpg "AISF - [4-(Acetylamino)phenyl]ImidodiSulfuryl")
## Advantages
- Bench stable, crystalline solid reagent used to synthesize fluorosulfates and sulfamoyl fluorides
- Non-hygroscopic
- No specialized equipment required – avoids the use of toxic sulfuryl fluoride gas
- Mild, room temperature conditions
- Excellent substrate scope
__Representative Procedure:__ To a one-dram vial containing the phenol or amine substrate (0.4 mmol) and AISF ([901243](https://www.sigmaaldrich.com/US/en/product/aldrich/901243); 134 mg, 0.48 mmol, 1.2 equiv.) was added tetrahydrofuran (2 mL) followed by 1,8-diazabicyclo \[5.4.0]undec-7-ene (132 µL, 0.88 mmol, 2.2 equiv.) over a period of 30 seconds. The reaction mixture was stirred at room temperature for 10 minutes and then diluted with ethyl acetate or ether and washed with either 0.5 N KHSO4 or 0.5 N HCl (2x) and brine (1x). The combined organic fraction was dried with anhydrous sodium sulfate and concentrated under reduced pressure. The crude residue was purified by silica gel flash chromatography.
#### Additional base and solvent combinations have also proved effective:
| | | | | | |
|-------|-------------|------------------------|------------|------------|---------|
| Entry | AISF equiv. | Base | Solvent | Time (min) | Yield |
| __1__ | __1.2__ | __DBU (2.2 equiv)__ | __THF__ | __10__ | __97%__ |
| __2__ | __1.2__ | __Cs2CO3 (1.2 equiv)__ | __THF__ | __20__ | __90%__ |
| __3__ | __1.2__ | __K2CO3 (3 equiv)__ | __DMSO__ | __60__ | __75%__ |
| __4__ | __1.2__ | __DBU (2.2 equiv)__ | __CH2CI2__ | __10__ | __96%__ |
| __5__ | __1.2__ | __Et3N__ | __DMSO__ | __10__ | __88%__ |
#### Application to single-flask transformations of phenols:
#### AISF can be applied to peptide functionalization under aqueous conditions:
*Special thanks to Chris am Ende (Pfizer) and the BioDuro Team for contributing this technology spotlight!*
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
### References
1\.
Zhou H, Mukherjee P, Liu R, Evrard E, Wang D, Humphrey JM, Butler TW, Hoth LR, Sperry JB, Sakata SK, et al. 2018. Introduction of a Crystalline, Shelf-Stable Reagent for the Synthesis of Sulfur(VI) Fluorides. Org. Lett.. 20(3):812-815. [https://doi.org/10.1021/acs.orglett.7b03950](https://doi.org/10.1021/acs.orglett.7b03950)
__Related Articles__
- [Acid Halogenation Reagents](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/fluorination/acid-halogenation)
- [AlPhos and \[(AlPhosPd)2•COD\] for Pd-Catalyzed Fluorination](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/fluorination/alphos)
- [Deoxyfluorination with 2-Pyridinesulfonyl Fluoride (PyFluor)](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/fluorination/deoxyfluorination-with-pyfluor)
- [Diethylaminosulfur Trifluoride (DAST)](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/fluorination/diethylaminosulfur)
- [Fluoroalkylation: Expansion of Togni Reagents](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/fluorination/fluoralkylation)
- [Fluorinated Azides: Click Chemistry Meets Fluorine](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/fluorination/fluorinated-azides)
- [PhenoFluor™ Deoxyfluorination Solution](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/fluorination/phenofluor)
- [Selectfluor™](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/fluorination/selectfluor)
- [View More](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search)
__Related Product Categories__
- [Caustic alkalis and bases](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/lab-chemicals/caustic-alkalis-and-bases)
Top
__Sign In To Continue__
Sign In__Don't Have An Account?__Register
Support[Customer Support](https://www.sigmaaldrich.com/US/en/support/customer-support)[Contact Us](https://www.sigmaaldrich.com/US/en/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Safety Data Sheets (SDS)](https://www.sigmaaldrich.com/US/en/documents-search?tab=sds)[Certificates (COA/COO)](https://www.sigmaaldrich.com/US/en/documents-search?tab=coa)[Quality & Regulatory](https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management)[Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps)[Webinars](https://www.sigmaaldrich.com/US/en/collections/webinars)
Orders[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)[Custom Products](https://www.sigmaaldrich.com/US/en/services/custom-products)[eCommerce Solutions](https://www.sigmaaldrich.com/US/en/life-science/ecommerce/ecommerce-solutions)
Company[About Us](https://www.sigmaaldrich.com/US/en/life-science/about-us)[Responsibility](https://www.sigmaaldrich.com/US/en/life-science/ssbi)[Events](https://www.sigmaaldrich.com/US/en/collections/events)[Press Releases](https://www.sigmaaldrich.com/US/en/collections/press)[Programs](https://www.sigmaaldrich.com/US/en/life-science/partnership-programs)[Careers](https://careers.merckgroup.com/global/en)[Offices](https://www.sigmaaldrich.com/US/en/collections/offices)
Social Media
[](https://www.linkedin.com/company/merck-life-science)
[](https://x.com/Merck_lifesci)
[](https://www.facebook.com/Merck.lifescience)
[](https://www.instagram.com/mercklifescience/)
[](https://www.youtube.com/@MerckLifeScience/featured)
MilliporeSigma
__Research. Development. Production.__
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
[](https://www.sigmaaldrich.com/US/en)
[Sigma-Aldrich® Solutions](https://www.sigmaaldrich.com/US/en/life-science/sigma-aldrich)[BioReliance® Solutions](https://www.sigmaaldrich.com/US/en/life-science/bioreliance)[Millipore® Solutions](https://www.sigmaaldrich.com/US/en/life-science/millipore)[SAFC® Solutions](https://www.sigmaaldrich.com/US/en/life-science/safc)[Milli-Q® Solutions](https://www.sigmaaldrich.com/US/en/life-science/milliq)[Supelco® Solutions](https://www.sigmaaldrich.com/US/en/life-science/supelco)
© 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies.
Reproduction of any materials from the site is strictly forbidden without permission.
[Site Use Terms](https://www.sigmaaldrich.com/US/en/life-science/legal/site-use-terms)|[Privacy Policy](https://www.sigmaaldrich.com/US/en/life-science/legal/privacy-statement)|[General Terms and Conditions of Sale](https://www.sigmaaldrich.com/US/en/life-science/legal/terms-and-conditions)|[Copyright Consent](https://www.sigmaaldrich.com/US/en/life-science/legal/copyright-consent) |[Site Map](https://www.sigmaaldrich.com/US/en/site-map)