MilliporeSigma
HomeWastewater & Process Water​ TestingAnalytical Method: Selenium in Water
Analytical Method: Selenium in Water
Instruments and Reagents
Product No. 109748 Spectroquant® Photometer NOVA 30 or
Product No. 109751 Spectroquant® Photometer NOVA 60
Product No. 114944 Rectangular cells 50 mm
Product No. 100264 Formic acid 98-100% for analysis
3,3‘-Diaminobenzidine tetrahydrochloride (CAS 868272-85-9), e. g. Sigma-Aldrich Art. 32750
Product No. 109992 TitriplexIII® solution 0.1 mol/l
Product No. 105432 Ammonia solution 25% for analysis
Product No. 108389 Toluene for gas chromatography
Product No. 109584 MColorpHastTM pH-indicator strips, pH 2.0 - 9.0
Product No. 106649 Sodium sulfate anhydrous for analysis
Product No. 116754 Water for analysis
Membrane filter with 0.45 μm pore diameter
Measuring Range
0.05 – 2.00 mg/L
Reagent 1
Fill 9.4 mL formic acid 100% in a calibrated or conformity-checked 100-mL volumetric flask. Make up to volume with water for analysis and mix.
Reagent 2
Dissolve 0.5 g 3,3’-diaminobenzidine tetrahydrochloride in 100 mL chilled water for analysis. Prepare freshly each day and store in an amber glass bottle.
Reagent 3
Fill 48 mL ammonia solution 25% for analysis in a calibrated or conformity-checked 100-mL volumetric flask. Make up to volume with water for analysis and mix.
Sample Preparation
The sample must be clear; if not, filter through a membrane filter with 0.45 μm pore diameter.
Analysis
Mix 30 mL nearly neutral sample solution with 2 mL reagent 1, 2 mL Titriplex III® solution 0.1 mol/l and 2 mL reagent 2. Adjust the pH value of the mixture with reagent 3 to 2.5 and set aside for 45 min at a dark place. Adjust the pH value of the mixture again with reagent 3 to 7.0, transfer into a 100-mL separation funnel, add 15 mL water for analysis and 7 mL toluene for analysis. Shake the mixture in the closed funnel for 1 min. Allow to separate and discharge the lower water phase. Fill the toluene extract in a test tube, add some anhydrous sodium sulfate and shake. Fill the clear, dried toluene extract in a 50 mm rectangular cell and measure at 445 nm against pure toluene.
Parameter List
Method number
----
Method designation
Selenium
Wavelength
445 nm
Citation
Se
Unit
mg/l
Resolution
0.01
Zero point (E0)
-0.037
Slope (E/C)
1.529
Meas. rang begin
0.05
Meas. range end
2.00
Reference cell
50 mm rect. cell
Calculation when measuring the absorbance value using NOVA photometers without programming the method in the memory of the instrument:
Concentration [mg/l] = Absorbance x 0.654 – 0.037
Calculation:
Result = measured value in mg/l Se4+
Statistical Data:
Standard deviation: 0.019 mg/l
Coefficient of variation: 2.9 %
Materials
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
Reference
1.
Fries J, Getrost H. 1975. Organische Reagenzien für die Spurenanalyse. Darmstadt: E. Merck.