MilliporeSigma
HomePharmacology & Drug Discovery ResearchFamciclovir Tablets USP Monograph Method Using a Purospher™ STAR RP-8 endcapped HPLC Column and UV Detection
Famciclovir Tablets USP Monograph Method Using a Purospher™ STAR RP-8 endcapped HPLC Column and UV Detection
Sonal Shinde, Application Specialist
Article from Analytix Reporter - Issue 6
Introduction
Famciclovir is an antiviral drug indicated for the treatment of herpes zoster, herpes simplex virus 2 (genital herpes), herpes labialis (cold sores), etc. It is a guanosine analogue, a prodrug form of penciclovir, and marketed by Novartis under the trade name Famvir. Generics are produced by TEVA and Mylan, among others. Purospher™ STAR RP-8 endcapped HPLC columns can be used to monitor organic impurities in tablet formulations following the new USP monograph for Famciclovir Tablets. The method suitability requirements are defined by the relative standard deviation (NMT 5.0% for Famciclovir standard solution) and the chromatographic resolution between propionyl famciclovir and 6-chloro famciclovir (NLT 1.2 using the system suitability solution).
Famciclovir
Experimental Conditions
HPLC Conditions 
ColumnPurospher™ STAR RP-8 endcapped (5 µm) 250x4.6 mm (1.51454)
Mobile Phase[A] 2.72 g/L of monobasic potassium phosphate in water. Adjust with 1 M phosphoric acid to a pH of 4.0 ±0.05.
 [B] Acetonitrile
GradientSee table
Flow Rate1 mL/min
Pressure Drop132-147 bar (1914-2131 psi)
DetectionUV @ 220 nm (analytical flow cell; 10 µL)
TemperaturesColumn: 50 °C; Autosampler: 8 °C
Injection Volume20 µL
Samples 
Standard Solution1 μg/mL of USP Famciclovir RS in Mobile Phase A (Figure 1)
SST Solution0.5 mg/mL of USP Famciclovir System Suitability Mixture RS in Mobile Phase A (Figure 2)
Test SolutionNominally 1 mg/mL of Famciclovir in Mobile Phase A prepared as follows. Transfer an amount equivalent to 250 mg of Famciclovir, from not less than (NLT) 10 finely powdered tablets, to a 250-mL volumetric flask. Add about 125 mL of mobile phase A and sonicate for 30 min with intermittent shaking. Dilute with mobile phase A to volume. (Figure 3)
 Other samples in monograph method (not shown here)
Peak ID Solution4 μg/mL of USP Famciclovir Related Compound A RS and 10 μg/mL of USP Famciclovir Related Compound B RS in Mobile Phase A
The method acceptance criteria are defined by the relative retention times for Famciclovir related compound A, Famciclovir related compound B, Famciclovir, 6-Chloro famciclovir, and Propionyl famciclovir and are about 0.2, 0.5, 1.0, 1.32, and 1.35, respectively. This application note illustrates with required analytical data that the method meets USP41-NF36 guidelines.
Gradient  
TimeA (%)B (%)
  095  5
507525
607525
6595  5
7595  5
Results
Chromatographic data – blank solution.
Figure 1. Chromatographic data – blank & standard solution.
Chromatographic data –  standard solution.
Chromatographic data - system suitability test (SST) solution.
Figure 2.Chromatographic data - system suitability test (SST) solution.
PeaksCompoundRetention Time (min)RRTResolutionTailing Factor
1Famciclovir Related compound B15.80.51-0.99
2Famciclovir30.81.0047.91.02
36-Chloro famciclovir40.71.3225.70.98
4Propionyl famciclovir41.41.34  1.70.98
Chromatographic data - test solution.
Figure 3. Chromatographic data - test solution.
PeaksCompoundRetention Time (min)RRTResolutionTailing Factor
1Famciclovir Related compound B15.80.51  00.99
2Famciclovir30.81.0047.51.02
36-Chloro famciclovir40.71.32  8.90.98
4Propionyl famciclovir41.41.34  7.70.98
Evaluation and verification
1. Specificity
Inject solution and determine the retention time of desired analyte in the presence of other components such as impurities and excipients.
PeaksCompoundRT (min)
1Famciclovir Related compound B15.8
2Famciclovir30.9
36-Chloro famciclovir40.7
4Propionyl famciclovir41.3
2. Standard Repeatability (1 ppm)
SampleArea Units
STD 1214,771
STD 2213,539
STD 3214,102
STD 4214,935
STD 5214,216
Mean214,313
Standard Deviation        559
RSD (%)         0.3
3.  Linearity, LOD & LOQ
Famciclovir Concentration (μg/mL)Area Units
0.5     59,951
1     98,532
5   478,367
12.51,190,770
252,360,140
403,757,032
504,687,957
605,618,958
757,007,616
LOD (ppm)            0.3
LOQ (ppm)            0.9
Concentration in ppm
Figure 4.Area / Concentration in ppm
Materials
Sorry, an unexpected error has occurred
Response not successful: Received status code 500