Merck
  • A chemical genetics analysis of the roles of bypass polymerase DinB and DNA repair protein AlkB in processing N2-alkylguanine lesions in vivo.
A chemical genetics analysis of the roles of bypass polymerase DinB and DNA repair protein AlkB in processing N2-alkylguanine lesions in vivo.
PloS one (2014-04-16)
Nidhi Shrivastav, Bogdan I Fedeles, Deyu Li, James C Delaney, Lauren E Frick, James J Foti, Graham C Walker, John M Essigmann
ZUSAMMENFASSUNG
DinB, the E. coli translesion synthesis polymerase, has been shown to bypass several N2-alkylguanine adducts in vitro, including N2-furfurylguanine, the structural analog of the DNA adduct formed by the antibacterial agent nitrofurazone. Recently, it was demonstrated that the Fe(II)- and α-ketoglutarate-dependent dioxygenase AlkB, a DNA repair enzyme, can dealkylate in vitro a series of N2-alkyguanines, including N2-furfurylguanine. The present study explored, head to head, the in vivo relative contributions of these two DNA maintenance pathways (replicative bypass vs. repair) as they processed a series of structurally varied, biologically relevant N2-alkylguanine lesions: N2-furfurylguanine (FF), 2-tetrahydrofuran-2-yl-methylguanine (HF), 2-methylguanine, and 2-ethylguanine. Each lesion was chemically synthesized and incorporated site-specifically into an M13 bacteriophage genome, which was then replicated in E. coli cells deficient or proficient for DinB and AlkB (4 strains in total). Biochemical tools were employed to analyze the relative replication efficiencies of the phage (a measure of the bypass efficiency of each lesion) and the base composition at the lesion site after replication (a measure of the mutagenesis profile of each lesion). The main findings were: 1) Among the lesions studied, the bulky FF and HF lesions proved to be strong replication blocks when introduced site-specifically on a single-stranded vector in DinB deficient cells. This toxic effect disappeared in the strains expressing physiological levels of DinB. 2) AlkB is known to repair N2-alkylguanine lesions in vitro; however, the presence of AlkB showed no relief from the replication blocks induced by FF and HF in vivo. 3) The mutagenic properties of the entire series of N2-alkyguanines adducts were investigated in vivo for the first time. None of the adducts were mutagenic under the conditions evaluated, regardless of the DinB or AlkB cellular status. Taken together, the data indicated that the cellular pathway to combat bulky N2-alkylguanine DNA adducts was DinB-dependent lesion bypass.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylendiamin, BioReagent, suitable for electrophoresis, ≥99.0%
Sigma-Aldrich
Acrylamid, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Phenol -Lösung, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylendiamin, BioReagent, Molecular Biology, ≥99% (GC)
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Acrylamid, Molecular Biology, ≥99% (HPLC)
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylendiamin, ≥99.5%, purified by redistillation
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Hydroxylapatit, nanopowder, <200 nm particle size (BET), ≥97%, synthetic
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Phenol -Lösung, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
3-Methyl-1-Butanol, reagent grade, 98%
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylendiamin, ReagentPlus®, 99%
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Isoamylalkohol, ≥98%, FG
Sigma-Aldrich
Verflüssigtes Phenol, ≥89.0%
Sigma-Aldrich
Acrylamid, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Acrylamid -Lösung, 40%, suitable for electrophoresis, sterile-filtered
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material