Sobre este item
Pular para
embalagem
pkg of 1 unit
fabricante/nome comercial
Milli-Q®
A × L × P
3.5 cm (1.4 in.) × 8.5 cm (3.3 in.) × 6.5 cm (2.5 in.)
Diâmetro
23 cm (9 in.)
compatibilidade
for use with Milli-Q® Reference
1 of 4
Este Item | ZEQ7C0NDC | ZFWATDET4 | TANKLK002 |
|---|---|---|---|
| compatibility for use with Milli-Q® Reference | compatibility for use with Milli-Q® EQ 7000 | compatibility for use with AFS®, for use with Elix® Advantage, for use with Elix® Essential UV, for use with Elix® Reference, for use with Milli-Q® Direct, for use with Milli-Q® Reference, for use with RiOs™ Essential | compatibility for use with Milli-Q® CLX, for use with Milli-Q® HR, for use with Milli-Q® HX |
| packaging pkg of 1 unit | packaging - | packaging pkg of 1 unit | packaging pkg of 1 unit |
| manufacturer/tradename Milli-Q® | manufacturer/tradename Milli-Q® | manufacturer/tradename - | manufacturer/tradename - |
| H × W × D 3.5 cm (1.4 in.) × 8.5 cm (3.3 in.) × 6.5 cm (2.5 in.) | H × W × D - | H × W × D - | H × W × D - |
| diam. 23 cm (9 in.) | diam. - | diam. - | diam. - |
Conteúdo relacionado
Feed Conductivity Cell datasheet