Merck
HS120639
Reagent Reservoir
capacity 10 mL, polystyrene, sterile; electron beam irradiated, pack of 100 (individually wrapped)
Sinônimo(s):
Solution Reservoir
Faça login para ver os preços organizacionais e de contrato.
Selecione um tamanho
100 EA
R$ 745,00
R$ 745,00
Verificar disponibilidade no carrinho
Sobre este item
NACRES:
NB.01
UNSPSC Code:
41121522
Pular para
Assistência Técnica
Precisa de ajuda? Nossa equipe de cientistas experientes está à sua disposição.
Estamos aqui para ajudar
Nome do produto
Reagent Reservoir, capacity 10 mL, polystyrene, sterile; electron beam irradiated, pack of 100 (individually wrapped)
material
V-bottom bottom
polystyrene
sterility
sterile; electron beam irradiated
packaging
pack of 100 (individually wrapped)
manufacturer/tradename
Heathrow Scientific 120639
capacity
10 mL
Comparar Itens Semelhantes
Ver Comparação Total
Mostrar diferenças
1 of 4
Este Item
HS120647HS120646HS120638
Reagent Reservoir capacity 10 mL, polystyrene, sterile; electron beam irradiated, pack of 100 (individually wrapped)
HS120639
Reagent Reservoir
Reagent Reservoir capacity 100 mL, polystyrene, sterile; electron beam irradiated, pack of 200 (packed as 5 ea)
HS120647
Reagent Reservoir
Reagent Reservoir capacity 50 mL, PVC, sterile; electron beam irradiated, pack of 100 (packed as 5 ea)
HS120646
Reagent Reservoir
Reagent Reservoir capacity 10 mL, polystyrene, non-sterile, pack of 100 (bulk packed)
HS120638
Reagent Reservoir
material
V-bottom bottom, polystyrene
material
V-bottom bottom, polystyrene
material
PVC , V-bottom bottom
material
V-bottom bottom, polystyrene
capacity
10 mL
capacity
100 mL
capacity
50 mL
capacity
10 mL
packaging
pack of 100 (individually wrapped)
packaging
pack of 200 (packed as 5 ea)
packaging
pack of 100 (packed as 5 ea)
packaging
pack of 100 (bulk packed)
sterility
sterile; electron beam irradiated
sterility
sterile; electron beam irradiated
sterility
sterile; electron beam irradiated
sterility
non-sterile
manufacturer/tradename
Heathrow Scientific 120639
manufacturer/tradename
Heathrow Scientific 120647
manufacturer/tradename
Heathrow Scientific 120646
manufacturer/tradename
Heathrow Scientific 120638
Features and Benefits
  • Trough design with a “v” shaped bottom helps ensure that all the reagent is accessible and can be easily aspirated
  • Thick wall construction and wide base adds stability and durability making the reservoir easy to handle without product deformation which results in less spillage
  • Reservoirs can be conveniently stacked with a unique nesting design that enables easy separation
  • Designed to stay flat on the benchtop and not warp
  • Sealed in easy-to-open bags. No scissors needed to open
  • Disposable
  • Design Patent Pending
General description
Reagent Reservoirs offer a fresh and modern design with enhanced features to streamline pipetting procedures. Graduation markingson the top edge and inner walls allows measurements of liquid to be easily identified and reduces waste of expensive reagents. Pour-off spouts design on all four corners enables easy spill-free pouring and reduces chance of dripping.
Escolha uma das versões mais recentes:
Certificados de análise (COA)