Merck
BMSIPS205020E
Benchmark OS-20L Overhead Stirrer
AC/DC input 220 V, Schuko plug
Synonym(e):
overhead electric stirrer, digital overhead stirrer
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Über diesen Artikel
NACRES:
NB.72
UNSPSC Code:
41103800
Fortfahren mit
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
Produktname
Benchmark OS-20L Overhead Stirrer, AC/DC input 220 V, Schuko plug
packaging
pack of 1 ea
manufacturer/tradename
Benchmark Scientific IPS2050-20-E
parameter
30-2200 rpm speed
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 4
Dieser Artikel
BMSIPS205040EZ767867BMSIPS2050PS1
manufacturer/tradename
Benchmark Scientific IPS2050-20-E
manufacturer/tradename
Benchmark Scientific IPS2050-40-E
manufacturer/tradename
IKA 4090000
manufacturer/tradename
Benchmark Scientific IPS2050-P-S1
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
parameter
30-2200 rpm speed
parameter
30-2200 rpm speed
parameter
0/4-530 rpm speed
parameter
-
Features and Benefits
  • LCD control panel
  • Includes complete stand assembly
  • Compact and quiet
  • Digital accuracy and convenience
General description
Benchmark′s Overhead Stirrers feature high torque and advanced digital microprocessor control for smooth mixing of low and medium viscosity samples. Speed and run time are accurately set and monitored on the LCD screen. They are supplied with a 4 arm stainless steel propeller, additional steel and PTFE accessories are available. A feed-through chuck holds accessory shafts firmly in place and is easily tightened and loosened without tools. The overhead stirrers are protected from damage by sensors that shut down operation in the case of high viscosity or obstruction, while a splash cover protects the connection mechanism from potential corrosion. Unlike most mixers, the Benchmark series is also supplied with a support stand and mounting hardware. The stirrer easily and securely attaches to the stand for compatibility with a variety of vessels, up to 24 inches high.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Questions
  1. What are the spec of this product? How tall and wide is it?
    1 answer
    1. The specifications, including the height and width, are as follows:
      Maximum Torque: 40Ncm
      Net Weight (Mixer): 2.6kg
      Maximum Viscosity: 10000mPas
      Speed Range: 30-2200 rpm
      Motor Type: DC brushless motor
      Chuck Diameter Range: 0.5-10mm
      Exterior Dims.: (WxDxH) 7.3 x 3.3 x 8.7 in.
      Ambient Operating Range: 5-40°C