Materiali
EPDM rubber device (ethylene propylene diene monomer)
PTFE
Livello qualitativo
Caratteristiche
hydrophobic (Surface)
Parametri
-40-170 °C temp. range (Continuous use)
-40-250 °C temp. range (Short term use)
Peso
0.18 kg (0.397 lb)
1 of 4
Questo articolo | NA12/22 | NA76/25 | NA12/25 |
|---|---|---|---|
| material EPDM rubber device (ethylene propylene diene monomer), PTFE | material EPDM rubber device (ethylene propylene diene monomer rubber), PTFE | material PTFE , PTFE device (polytetrafluoroethylene) | material PTFE , PTFE device (polytetrafluoroethylene), valve (Diaphragm, PTFE) |
| weight 0.18 kg (0.397 lb) | weight 0.01 kg (0.022 lb) | weight 0.16 kg (0.353 lb) | weight 0.01 kg (0.022 lb) |
| feature hydrophobic (Surface) | feature hydrophobic (Surface) | feature hydrophobic (Surface) | feature hydrophobic (Surface) |
| parameter -40-170 °C temp. range (Continuous use) | parameter -30-140 °C temp. range (Short term use), -6-(-1) bar pressure range | parameter -40-170 °C temp. range (Continuous use), -40-250 °C temp. range (Short term use) | parameter -40-170 °C temp. range (Continous use), -40-250 °C temp. range (Short term use) |
| Quality Level SP2 | Quality Level SP2 | Quality Level SP2 | Quality Level SP2 |
Applicazioni
Confezionamento
Filtri attivi
Contatta l'Assistenza Tecnica