Troubleshooting PCR and RT-PCR Amplification
Many of the common problems with PCR and RT-PCR are identified during agarose gel electrophoresis of the reaction products. These include the absence of the expected amplification product, the presence of nonspecific products, excessive smearing, and the presence of a “primer dimer” band. A summary of the possible causes and solutions for these problems is provided below.
| Problem | Possible cause | Solution |
|---|---|---|
| No bands on gel | The thermal cycler was not functioning properly | If a positive control was not prepared, consider testing reagents in a control reaction with template and primers previously shown to function properly. |
| Insufficient template or too much template was added | Titrate the amount of template required for your system. | |
| For DNA: Template quality was poor or contains inhibitors | Impure DNA can fail to amplify properly. Use freshly prepared DNA or isolate template by another method. | |
| Primer concentration was too low or was unbalanced | Make sure primer concentration is within recommended range and that concentration of both PCR primers is the same. | |
| Nucleases were introduced | Prepare new template. Be sure to follow precautions against introducing nucleases. This is particularly important when the starting material is RNA. | |
| For RNA: Template is in buffer containing EDTA | EDTA present in the RNA solution will chelate Mg2+ required for the RT-PCR and may lead to suboptimal results. To compensate, add additional Mg2+ when assembling the reactions. Refer to product instructions for details. | |
| Reagents were not thawed properly before use | Thaw and thoroughly mix all reagents before setting up reactions. | |
| Primer annealing temperature was too high | Decrease annealing temperature in 2 °C increments. | |
| Denaturing temperature was suboptimal | Try increasing the temperature or duration time of the initial denaturation step. This is especially useful for GC rich templates. | |
| Extension time was too short | Increase extension times. This can be especially important for long PCR. | |
| Thermal cycler was not at correct temperature | Repair/calibrate the thermal cycler or use a different machine. | |
| Extra, nonspecific bands on gel | Primers hybridized to a secondary site on the template | Design new primers that are less specific for the secondary site. Increase the annealing temperature by increments of 2 °C to 5 °C. |
| DNA contamination was introduced in primers or buffers | Run a negative control reaction (no template). Prepare fresh materials if contamination is detected. | |
| For RNA: Template is contaminated with DNA | Perform a DNase step and completely inactivate/ remove DNase. Alternatively, prepare RNA using a different method. | |
| Too much template was added | Titrate the amount of template for your system. | |
| Primer concentration was too high | Titrate the amount of primers for your system. | |
| Thermal cycler was not at correct temperature | Repair/calibrate the thermal cycler or use a different machine. | |
| Excessive smearing of amplified DNA | Too much template was added | Titrate the amount of template for your system. |
| Too many cycles were performed | Reduce the number of cycles below 35. | |
| Thermal cycling conditions were not optimal for your thermal cycler | Optimize conditions based on manufacturer’s recommendations. | |
| The annealing temperature was too low | When using Ready-To-Go Beads, re-optimization of the annealing temperature might be required. Increase the annealing temperature by increments of 2 °C to 5 °C. | |
| Primer concentration was too high | Titrate the amount of primers for your system. | |
| Extension time was too long | Decrease extension time in small increments. | |
| For DNA: Template quality was low | Impure DNA can fail to amplify properly. Use freshly prepared DNA or isolate template by another method. | |
| Too much DNA was loaded on gel | Re-run with less sample. | |
| Primer dimers visible | Too much primer was added/ primer concentration too high | Titrate the amount of primers for your system. |
| Primers possess complementary overlapping sequence | Design primers to avoid self-complementary internal sequences. Increase annealing temperature. Use illustra™ Hot Start Mix PCR products. |
Materials
Si è verificato un errore inatteso.
Response not successful: Received status code 500